Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591196_m1
          See other MUTYH GE Assays ›
          Gene Symbol:
          Mutyh
          Gene Name:
          mutY homolog
          Gene Aliases:
          Myh
          Chromosome Location:
          Chr.5: 135663332 - 135675348 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591196_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mutyh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133316.1 NP_579850.1 15 - 16 1481 77
          XM_006238626.2 XP_006238688.1 15 - 16 1509 77
          XM_006238627.2 XP_006238689.1 14 - 15 1406 77
          XM_008763987.1 XP_008762209.1 13 - 14 1467 77
          GenBank mRNA AF478683.1 - 15 - 16 1481 77
          BC128728.1 - 12 - 13 1828 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001275.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mutyh

          Panther Classification:

          Molecular Function -

          DNA glycosylase

          Gene Ontology Categories:

          Function(s) Process(es)

          base-excision repair
          response to oxidative stress
          DNA N-glycosylase activity
          MutLalpha complex binding
          MutLbeta complex binding
          MutSalpha complex binding
          MutSbeta complex binding
          metal ion binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722447 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007332 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007332 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P505729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74288 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52678 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871323 - -
          Affymetrix Rat Gene 1.0 ST Array 10871328 - -
          Affymetrix Rat Gene 1.0 ST Array 10871326 - -
          Affymetrix Rat Gene 1.0 ST Array 10871333 - -
          Affymetrix Rat Gene 1.0 ST Array 10871327 - -
          Affymetrix Rat Gene 1.0 ST Array 10871324 - -
          Affymetrix Rat Gene 1.0 ST Array 10871322 - -
          Affymetrix Rat Gene 1.0 ST Array 10871321 - -
          Affymetrix Rat Gene 1.0 ST Array 10871335 - -
          Affymetrix Rat Gene 1.0 ST Array 10871334 - -
          Affymetrix Rat Gene 1.0 ST Array 10871330 - -
          Affymetrix Rat Gene 1.0 ST Array 10871332 - -
          Affymetrix Rat Gene 1.0 ST Array 10871331 - -
          Affymetrix Rat Gene 1.0 ST Array 10871329 - -
          Affymetrix Rat Gene 1.0 ST Array 10871336 - -
          Affymetrix Rat Gene 1.0 ST Array 10871325 - -
          Affymetrix Rat Gene 1.1 ST Array 10871319 - -
          Rat Genome 230 2.0 Array 1368787_at - -
          Rat Genome 230 2.0 Array 1368787_PM_at - -
          Affymetrix HT Rat Focus Array 1368787_at - -
          Affymetrix HT Rat Focus Array 1368787_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500008902.rn.1 TC0500001275.rn.1 RTA TC
          Rat Expression Set 230 1368787_at - -
          Rat Expression Set 230 1368787_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871319 - -
          Rat Genome U34 Set rc_AI043911_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21744924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361131 - -
          RatRef-12 Expression BeadChip ILMN_66002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline