Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592040_m1
          See other INPP5J GE Assays ›
          Gene Symbol:
          Inpp5j
          Gene Name:
          inositol polyphosphate-5-phosphatase J
          Gene Aliases:
          Inpp, Pib5pa, Pipp
          Chromosome Location:
          Chr.14: 83730861 - 83742011 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592040_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Inpp5j
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133562.1 NP_598246.1 9 - 10 2218 70
          XM_006251155.2 XP_006251217.1 9 - 10 2239 70
          XM_008770248.1 XP_008768470.1 9 - 10 2260 70
          GenBank mRNA AB032551.1 - 9 - 10 2218 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002128.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Inpp5j

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of neuron projection development
          dephosphorylation
          negative regulation of microtubule polymerization
          negative regulation of peptidyl-serine phosphorylation
          phosphatidylinositol dephosphorylation
          inositol-polyphosphate 5-phosphatase activity
          SH3 domain binding
          phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
          inositol-1,4,5-trisphosphate 5-phosphatase activity
          inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P201178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59400 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P201178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P201178 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P201178 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P201178 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700325265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61490 - -
          Agilent Rat cDNA Microarray (G4105A) 700325265 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777959 - -
          Affymetrix Rat Gene 1.0 ST Array 10777969 - -
          Affymetrix Rat Gene 1.0 ST Array 10777968 - -
          Affymetrix Rat Gene 1.0 ST Array 10777962 - -
          Affymetrix Rat Gene 1.0 ST Array 10777961 - -
          Affymetrix Rat Gene 1.0 ST Array 10777964 - -
          Affymetrix Rat Gene 1.0 ST Array 10777966 - -
          Affymetrix Rat Gene 1.0 ST Array 10777960 - -
          Affymetrix Rat Gene 1.0 ST Array 10777967 - -
          Affymetrix Rat Gene 1.0 ST Array 10777971 - -
          Affymetrix Rat Gene 1.0 ST Array 10777963 - -
          Affymetrix Rat Gene 1.0 ST Array 10777965 - -
          Affymetrix Rat Gene 1.0 ST Array 10777970 - -
          Affymetrix Rat Gene 1.1 ST Array 10777958 - -
          Rat Genome 230 2.0 Array 1368119_PM_at - -
          Rat Genome 230 2.0 Array 1368119_at - -
          Affymetrix HT Rat Focus Array 1368119_at - -
          Affymetrix HT Rat Focus Array 1368119_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400015628.rn.1 TC1400002128.rn.1 RTA TC
          Rat Expression Set 230 1368119_PM_at - -
          Rat Expression Set 230 1368119_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777958 - -
          Rat Genome U34 Set rc_AI169384_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21875432 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349574 - -
          RatRef-12 Expression BeadChip ILMN_54155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline