Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593473_m1
          See other ERO1L GE Assays ›
          Gene Symbol:
          Ero1l
          Gene Name:
          ERO1-like (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 19619947 - 19655381 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593473_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ero1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138528.1 NP_612537.1 1 - 2 126 129
          GenBank mRNA AF489855.1 - 1 - 2 113 129
          AY071924.1 - 1 - 2 126 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001440.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ero1l

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          4-hydroxyproline metabolic process
          protein maturation by protein folding
          extracellular matrix organization
          endoplasmic reticulum unfolded protein response
          response to endoplasmic reticulum stress
          cell redox homeostasis
          brown fat cell differentiation
          chaperone mediated protein folding requiring cofactor
          release of sequestered calcium ion into cytosol
          oxidation-reduction process
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          protein disulfide isomerase activity
          protein disulfide oxidoreductase activity
          oxidoreductase activity
          oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P212826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99380 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212826 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P212826 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212826 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782844 - -
          Affymetrix Rat Gene 1.0 ST Array 10782827 - -
          Affymetrix Rat Gene 1.0 ST Array 10782839 - -
          Affymetrix Rat Gene 1.0 ST Array 10782829 - -
          Affymetrix Rat Gene 1.0 ST Array 10782840 - -
          Affymetrix Rat Gene 1.0 ST Array 10782835 - -
          Affymetrix Rat Gene 1.0 ST Array 10782841 - -
          Affymetrix Rat Gene 1.0 ST Array 10782832 - -
          Affymetrix Rat Gene 1.0 ST Array 10782838 - -
          Affymetrix Rat Gene 1.0 ST Array 10782831 - -
          Affymetrix Rat Gene 1.0 ST Array 10782843 - -
          Affymetrix Rat Gene 1.0 ST Array 10782828 - -
          Affymetrix Rat Gene 1.0 ST Array 10782834 - -
          Affymetrix Rat Gene 1.0 ST Array 10782830 - -
          Affymetrix Rat Gene 1.0 ST Array 10782837 - -
          Affymetrix Rat Gene 1.0 ST Array 10782833 - -
          Affymetrix Rat Gene 1.0 ST Array 10782845 - -
          Affymetrix Rat Gene 1.0 ST Array 10782842 - -
          Affymetrix Rat Gene 1.0 ST Array 10782836 - -
          Affymetrix Rat Gene 1.1 ST Array 10782826 - -
          Rat Genome 230 2.0 Array 1387964_PM_a_at - -
          Rat Genome 230 2.0 Array 1389297_at - -
          Rat Genome 230 2.0 Array 1387964_a_at - -
          Rat Genome 230 2.0 Array 1389297_PM_at - -
          Affymetrix HT Rat Focus Array 1389297_at - -
          Affymetrix HT Rat Focus Array 1389297_PM_at - -
          Affymetrix HT Rat Focus Array 1387964_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387964_a_at - -
          Affymetrix Rat Clariom S Assay PSR1500009182.rn.1 TC1500001440.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009184.rn.1 TC1500001440.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009183.rn.1 TC1500001440.rn.1 RTA TC
          Rat Expression Set 230 1389297_at - -
          Rat Expression Set 230 1387964_PM_a_at - -
          Rat Expression Set 230 1389297_PM_at - -
          Rat Expression Set 230 1387964_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782826 - -
          Rat Genome U34 Set rc_AI146215_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20702966 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376500 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline