Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594078_m1
          See other IFNG GE Assays ›
          Gene Symbol:
          Ifng
          Gene Name:
          interferon gamma
          Gene Aliases:
          IFNG2
          Chromosome Location:
          Chr.7: 61337383 - 61341419 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594078_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifng
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138880.2 NP_620235.1 3 - 4 373 91
          GenBank mRNA AF010466.1 - 3 - 4 373 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000775.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Tumor Metastasis Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifng

          Panther Classification:

          Molecular Function -

          cytokine interferon superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          negative regulation of transcription from RNA polymerase II promoter
          neutrophil apoptotic process
          regulation of the force of heart contraction
          adaptive immune response
          CD8-positive, alpha-beta T cell differentiation involved in immune response
          regulation of transcription, DNA-templated
          apoptotic process
          inflammatory cell apoptotic process
          humoral immune response
          cell cycle arrest
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to virus
          antigen processing and presentation
          neutrophil chemotaxis
          negative regulation of epithelial cell differentiation
          endoplasmic reticulum unfolded protein response
          negative regulation of myelination
          positive regulation of synaptic transmission, cholinergic
          negative regulation of interleukin-17 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-23 production
          positive regulation of tumor necrosis factor production
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          positive regulation of smooth muscle cell apoptotic process
          positive regulation of T cell proliferation
          response to drug
          positive regulation of tyrosine phosphorylation of Stat1 protein
          defense response to bacterium
          defense response to protozoan
          negative regulation of growth of symbiont in host
          negative regulation of growth of symbiont involved in interaction with host
          positive regulation of chemokine biosynthetic process
          positive regulation of interleukin-12 biosynthetic process
          positive regulation of MHC class II biosynthetic process
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of neuron differentiation
          positive regulation of osteoclast differentiation
          positive regulation of cell adhesion
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgG isotypes
          negative regulation of smooth muscle cell proliferation
          positive regulation of interleukin-1 beta secretion
          regulation of immune response
          regulation of insulin secretion
          sensory perception of mechanical stimulus
          positive regulation of membrane protein ectodomain proteolysis
          defense response to virus
          positive regulation of killing of cells of other organism
          positive regulation of fructose 1,6-bisphosphate 1-phosphatase activity
          positive regulation of fructose 1,6-bisphosphate metabolic process
          positive regulation of vitamin D biosynthetic process
          positive regulation of calcidiol 1-monooxygenase activity
          cellular response to lipopolysaccharide
          cellular response to interleukin-18
          extrinsic apoptotic signaling pathway
          regulation of neuronal action potential
          positive regulation of tumor necrosis factor (ligand) superfamily member 11 production
          regulation of hepatocyte proliferation
          cytokine activity
          interferon-gamma receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0700005965.rn.1 TC0700000775.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline