Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594398_m1
          See other FNBP1 GE Assays ›
          Gene Symbol:
          Fnbp1
          Gene Name:
          formin binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 9865657 - 9936333 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594398_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fnbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138914.1 NP_620269.1 140 71
          XM_006233929.2 XP_006233991.1 2 - 3 1536 71
          GenBank mRNA AB073208.1 - 140 71
          AB126168.1 - 140 71
          AB126169.1 - 140 71
          AB126170.1 - 140 71
          AB126171.1 - 140 71
          AB126172.1 - 140 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002845.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fnbp1

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          nervous system development
          cell differentiation
          protein binding
          lipid binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P174571 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868698 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78135 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13339 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P174571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P174571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13339 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P174571 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13339 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P430989 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13895 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19910 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17732 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24114 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69887 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844260 - -
          Affymetrix Rat Gene 1.0 ST Array 10844255 - -
          Affymetrix Rat Gene 1.0 ST Array 10844261 - -
          Affymetrix Rat Gene 1.0 ST Array 10844257 - -
          Affymetrix Rat Gene 1.0 ST Array 10844262 - -
          Affymetrix Rat Gene 1.0 ST Array 10844254 - -
          Affymetrix Rat Gene 1.0 ST Array 10844264 - -
          Affymetrix Rat Gene 1.0 ST Array 10844256 - -
          Affymetrix Rat Gene 1.0 ST Array 10844253 - -
          Affymetrix Rat Gene 1.0 ST Array 10844266 - -
          Affymetrix Rat Gene 1.0 ST Array 10844265 - -
          Affymetrix Rat Gene 1.0 ST Array 10844259 - -
          Affymetrix Rat Gene 1.0 ST Array 10844263 - -
          Affymetrix Rat Gene 1.0 ST Array 10844258 - -
          Affymetrix Rat Gene 1.0 ST Array 10844252 - -
          Affymetrix Rat Gene 1.1 ST Array 10844251 - -
          Rat Genome 230 2.0 Array 1375406_PM_s_at - -
          Rat Genome 230 2.0 Array 1390682_PM_at - -
          Rat Genome 230 2.0 Array 1390682_at - -
          Rat Genome 230 2.0 Array 1372825_at - -
          Rat Genome 230 2.0 Array 1377342_s_at - -
          Rat Genome 230 2.0 Array 1377342_PM_s_at - -
          Rat Genome 230 2.0 Array 1369471_at - -
          Rat Genome 230 2.0 Array 1376784_PM_at - -
          Rat Genome 230 2.0 Array 1372825_PM_at - -
          Rat Genome 230 2.0 Array 1376784_at - -
          Rat Genome 230 2.0 Array 1375406_s_at - -
          Rat Genome 230 2.0 Array 1369471_PM_at - -
          Affymetrix HT Rat Focus Array 1372825_at - -
          Affymetrix HT Rat Focus Array 1375406_PM_s_at - -
          Affymetrix HT Rat Focus Array 1376784_PM_at - -
          Affymetrix HT Rat Focus Array 1369471_at - -
          Affymetrix HT Rat Focus Array 1390682_at - -
          Affymetrix HT Rat Focus Array 1377342_s_at - -
          Affymetrix HT Rat Focus Array 1375406_s_at - -
          Affymetrix HT Rat Focus Array 1376784_at - -
          Affymetrix HT Rat Focus Array 1372825_PM_at - -
          Affymetrix HT Rat Focus Array 1390682_PM_at - -
          Affymetrix HT Rat Focus Array 1377342_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369471_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300021759.rn.1 TC0300002845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300021760.rn.1 TC0300002845.rn.1 RTA TC
          Rat Expression Set 230 1372825_at - -
          Rat Expression Set 230 1369471_PM_at - -
          Rat Expression Set 230 1376784_PM_at - -
          Rat Expression Set 230 1376784_at - -
          Rat Expression Set 230 1377342_s_at - -
          Rat Expression Set 230 1377342_PM_s_at - -
          Rat Expression Set 230 1369471_at - -
          Rat Expression Set 230 1390682_PM_at - -
          Rat Expression Set 230 1390682_at - -
          Rat Expression Set 230 1375406_s_at - -
          Rat Expression Set 230 1372825_PM_at - -
          Rat Expression Set 230 1375406_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844251 - -
          Rat Genome U34 Set rc_AI011679_at - -
          Rat Genome U34 Set rc_AI012469_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21419540 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373297 - -
          RatRef-12 Expression BeadChip ILMN_53717 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline