Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594462_m1
          See other EML2 GE Assays ›
          Gene Symbol:
          Eml2
          Gene Name:
          echinoderm microtubule associated protein like 2
          Gene Aliases:
          Emap, Emap-2, Eml1
          Chromosome Location:
          Chr.1: 80087684 - 80118784 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594462_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eml2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138921.1 NP_620276.1 21 - 22 2300 74
          GenBank mRNA AF335571.1 - 21 - 22 2300 74
          BC062038.1 - 18 - 19 1842 5598497 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000991.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eml2

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          regulation of microtubule nucleation
          negative regulation of microtubule polymerization
          catalytic activity
          receptor binding
          microtubule binding
          protein C-terminus binding
          tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P290357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165717 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290357 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290357 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P290357 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P290357 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39112 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37533 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704766 - -
          Affymetrix Rat Gene 1.0 ST Array 10704777 - -
          Affymetrix Rat Gene 1.0 ST Array 10704763 - -
          Affymetrix Rat Gene 1.0 ST Array 10704778 - -
          Affymetrix Rat Gene 1.0 ST Array 10704762 - -
          Affymetrix Rat Gene 1.0 ST Array 10704771 - -
          Affymetrix Rat Gene 1.0 ST Array 10704760 - -
          Affymetrix Rat Gene 1.0 ST Array 10704765 - -
          Affymetrix Rat Gene 1.0 ST Array 10704774 - -
          Affymetrix Rat Gene 1.0 ST Array 10704767 - -
          Affymetrix Rat Gene 1.0 ST Array 10704783 - -
          Affymetrix Rat Gene 1.0 ST Array 10704775 - -
          Affymetrix Rat Gene 1.0 ST Array 10704779 - -
          Affymetrix Rat Gene 1.0 ST Array 10704770 - -
          Affymetrix Rat Gene 1.0 ST Array 10704768 - -
          Affymetrix Rat Gene 1.0 ST Array 10704772 - -
          Affymetrix Rat Gene 1.0 ST Array 10704782 - -
          Affymetrix Rat Gene 1.0 ST Array 10704764 - -
          Affymetrix Rat Gene 1.0 ST Array 10704759 - -
          Affymetrix Rat Gene 1.0 ST Array 10704776 - -
          Affymetrix Rat Gene 1.0 ST Array 10704781 - -
          Affymetrix Rat Gene 1.0 ST Array 10704769 - -
          Affymetrix Rat Gene 1.0 ST Array 10704761 - -
          Affymetrix Rat Gene 1.0 ST Array 10704773 - -
          Affymetrix Rat Gene 1.0 ST Array 10704780 - -
          Affymetrix Rat Gene 1.1 ST Array 10704758 - -
          Rat Genome 230 2.0 Array 1370544_PM_at - -
          Rat Genome 230 2.0 Array 1370544_at - -
          Affymetrix HT Rat Focus Array 1370544_PM_at - -
          Affymetrix HT Rat Focus Array 1370544_at - -
          Affymetrix Rat Clariom S Assay PSR0100006957.rn.1 TC0100000991.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006958.rn.1 TC0100000991.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006959.rn.1 TC0100000991.rn.1 RTA TC
          Rat Expression Set 230 1370544_PM_at - -
          Rat Expression Set 230 1370544_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704758 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21471076 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352930 - -
          RatRef-12 Expression BeadChip ILMN_54589 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline