Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596306_m1
          See other ST18 GE Assays ›
          Gene Symbol:
          St18
          Gene Name:
          suppression of tumorigenicity 18
          Gene Aliases:
          Nzf3, r-MyT3
          Chromosome Location:
          Chr.5: 12437617 - 12565774 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596306_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: St18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153310.2 NP_695222.2 18 - 19 2799 115
          XM_008763489.1 XP_008761711.1 18 - 19 3354 115
          XM_008763490.1 XP_008761712.1 19 - 20 4015 115
          XM_008763491.1 XP_008761713.1 18 - 19 3785 115
          XM_008763492.1 XP_008761714.1 18 - 19 3859 115
          XM_008763493.1 XP_008761715.1 17 - 18 2717 115
          XM_008763494.1 XP_008761716.1 14 - 15 2501 115
          GenBank mRNA AF031942.1 - 15 - 16 2623 115
          U67080.1 - 18 - 19 2809 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002206.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene St18

          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158005 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105713 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105713 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105713 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292665 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94039 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29387 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82342 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875170 - -
          Affymetrix Rat Gene 1.0 ST Array 10875161 - -
          Affymetrix Rat Gene 1.0 ST Array 10875176 - -
          Affymetrix Rat Gene 1.0 ST Array 10875166 - -
          Affymetrix Rat Gene 1.0 ST Array 10875156 - -
          Affymetrix Rat Gene 1.0 ST Array 10875162 - -
          Affymetrix Rat Gene 1.0 ST Array 10875171 - -
          Affymetrix Rat Gene 1.0 ST Array 10875155 - -
          Affymetrix Rat Gene 1.0 ST Array 10875177 - -
          Affymetrix Rat Gene 1.0 ST Array 10875164 - -
          Affymetrix Rat Gene 1.0 ST Array 10875168 - -
          Affymetrix Rat Gene 1.0 ST Array 10875160 - -
          Affymetrix Rat Gene 1.0 ST Array 10875167 - -
          Affymetrix Rat Gene 1.0 ST Array 10875169 - -
          Affymetrix Rat Gene 1.0 ST Array 10875175 - -
          Affymetrix Rat Gene 1.0 ST Array 10875159 - -
          Affymetrix Rat Gene 1.0 ST Array 10875157 - -
          Affymetrix Rat Gene 1.0 ST Array 10875173 - -
          Affymetrix Rat Gene 1.0 ST Array 10875165 - -
          Affymetrix Rat Gene 1.0 ST Array 10875174 - -
          Affymetrix Rat Gene 1.0 ST Array 10875172 - -
          Affymetrix Rat Gene 1.0 ST Array 10875158 - -
          Affymetrix Rat Gene 1.0 ST Array 10875163 - -
          Affymetrix Rat Gene 1.1 ST Array 10875154 - -
          Rat Genome 230 2.0 Array 1370658_a_at - -
          Rat Genome 230 2.0 Array 1370658_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370658_a_at - -
          Affymetrix HT Rat Focus Array 1370658_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500016790.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016795.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016798.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016802.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016806.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016789.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016805.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016788.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016791.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016793.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016801.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016785.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016786.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016794.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016796.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016800.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016803.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016792.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016799.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016804.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016797.rn.1 TC0500002206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500016807.rn.1 TC0500002206.rn.1 RTA TC
          Rat Expression Set 230 1370658_a_at - -
          Rat Expression Set 230 1370658_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875154 - -
          Rat Genome U34 Set U67080_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22343328 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359803 - -
          RatRef-12 Expression BeadChip ILMN_62454 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline