Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597407_m1
          See other TUBB5 GE Assays ›
          Gene Symbol:
          Tubb5
          Gene Name:
          tubulin, beta 5 class I
          Gene Aliases:
          Tubb
          Chromosome Location:
          Chr.20: 3422448 - 3426420 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597407_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173102.2 NP_775125.1 2 - 3 359 68
          GenBank mRNA AB011679.1 - 2 - 3 318 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000108.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tubb5

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          biological_process
          metabolic process
          cellular process
          spindle assembly
          protein polymerization
          GTPase activity
          structural constituent of cytoskeleton
          protein binding
          GTP binding
          protein domain specific binding
          ubiquitin protein ligase binding
          protein complex binding
          GTPase activating protein binding
          MHC class I protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P172814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445780 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P172814 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P172814 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P172814 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P172814 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700182562 - -
          Agilent Rat cDNA Microarray (G4105A) 700182562 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10827910 - -
          Affymetrix Rat Gene 1.0 ST Array 10827913 - -
          Affymetrix Rat Gene 1.0 ST Array 10827915 - -
          Affymetrix Rat Gene 1.0 ST Array 10827911 - -
          Affymetrix Rat Gene 1.0 ST Array 10827907 - -
          Affymetrix Rat Gene 1.0 ST Array 10827909 - -
          Affymetrix Rat Gene 1.0 ST Array 10827908 - -
          Affymetrix Rat Gene 1.0 ST Array 10827914 - -
          Affymetrix Rat Gene 1.0 ST Array 10827912 - -
          Affymetrix Rat Gene 1.1 ST Array 10827906 - -
          Rat Genome 230 2.0 Array 1387892_PM_at - -
          Rat Genome 230 2.0 Array 1387892_at - -
          Rat Genome 230 2.0 Array 1370290_at - -
          Rat Genome 230 2.0 Array 1370290_PM_at - -
          Affymetrix HT Rat Focus Array 1370290_at - -
          Affymetrix HT Rat Focus Array 1387892_at - -
          Affymetrix HT Rat Focus Array 1370290_PM_at - -
          Affymetrix HT Rat Focus Array 1387892_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000000719.rn.1 TC2000000108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000723.rn.1 TC2000000108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000717.rn.1 TC2000000108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000721.rn.1 TC2000000108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000716.rn.1 TC2000000108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000000722.rn.1 TC2000000108.rn.1 RTA TC
          Rat Expression Set 230 1370290_at - -
          Rat Expression Set 230 1387892_at - -
          Rat Expression Set 230 1387892_PM_at - -
          Rat Expression Set 230 1370290_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10827906 - -
          Rat Genome U34 Set AB011679_at - -
          Rat Genome U34 Set rc_AA860030_s_at - -
          Rat Genome U34 Set rc_AA943532_s_at - -
          Rat Genome U34 Set rc_AA850450_f_at - -
          Rat Genome U34 Set rc_AI229707_s_at - -
          Rat Genome U34 Set rc_AA860030_s_at - -
          Rat Genome U34 Set rc_AA943532_s_at - -
          Rat Genome U34 Set AB011679_at - -
          Rat Genome U34 Set rc_AI229707_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21685981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370544 - -
          RatRef-12 Expression BeadChip ILMN_66560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline