Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597481_m1
          See other SGPL1 GE Assays ›
          Gene Symbol:
          Sgpl1
          Gene Name:
          sphingosine-1-phosphate lyase 1
          Gene Aliases:
          S1PL, Spgl1, spl
          Chromosome Location:
          Chr.20: 30699936 - 30769178 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597481_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sgpl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173116.1 NP_775139.1 3 - 4 317 109
          XM_008772870.1 XP_008771092.1 3 - 4 256 109
          XM_008772871.1 XP_008771093.1 3 - 4 266 109
          XM_008772872.1 XP_008771094.1 2 - 3 237 109
          XM_008772873.1 XP_008771095.1 3 - 4 324 109
          GenBank mRNA AJ512838.1 - 3 - 4 317 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001536.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sgpl1

          Gene Ontology Categories:

          Function(s) Process(es)

          luteinization
          vasculogenesis
          kidney development
          fatty acid metabolic process
          ceramide metabolic process
          nitrogen compound metabolic process
          spermatogenesis
          androgen metabolic process
          estrogen metabolic process
          female gonad development
          post-embryonic development
          fibroblast migration
          hemopoiesis
          sphingolipid catabolic process
          Leydig cell differentiation
          regulation of multicellular organism growth
          platelet-derived growth factor receptor signaling pathway
          skeletal system morphogenesis
          palate development
          face morphogenesis
          apoptotic signaling pathway
          sphinganine-1-phosphate aldolase activity
          carboxy-lyase activity
          pyridoxal phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158336 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168554 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23777 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063463 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063463 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063463 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P400211 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44880 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37678 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833036 - -
          Affymetrix Rat Gene 1.0 ST Array 10833033 - -
          Affymetrix Rat Gene 1.0 ST Array 10833041 - -
          Affymetrix Rat Gene 1.0 ST Array 10833039 - -
          Affymetrix Rat Gene 1.0 ST Array 10833034 - -
          Affymetrix Rat Gene 1.0 ST Array 10833037 - -
          Affymetrix Rat Gene 1.0 ST Array 10833035 - -
          Affymetrix Rat Gene 1.0 ST Array 10833045 - -
          Affymetrix Rat Gene 1.0 ST Array 10833032 - -
          Affymetrix Rat Gene 1.0 ST Array 10833046 - -
          Affymetrix Rat Gene 1.0 ST Array 10833043 - -
          Affymetrix Rat Gene 1.0 ST Array 10833040 - -
          Affymetrix Rat Gene 1.0 ST Array 10833044 - -
          Affymetrix Rat Gene 1.0 ST Array 10833038 - -
          Affymetrix Rat Gene 1.0 ST Array 10833042 - -
          Affymetrix Rat Gene 1.1 ST Array 10833031 - -
          Rat Genome 230 2.0 Array 1382843_at - -
          Rat Genome 230 2.0 Array 1382843_PM_at - -
          Affymetrix HT Rat Focus Array 1382843_PM_at - -
          Affymetrix HT Rat Focus Array 1382843_at - -
          Affymetrix Rat Clariom S Assay PSR2000012499.rn.1 TC2000001536.rn.1 RTA TC
          Rat Expression Set 230 1382843_at - -
          Rat Expression Set 230 1382843_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833031 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21577869 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352496 - -
          RatRef-12 Expression BeadChip ILMN_57727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0