Search
Search
| Gene Symbol: Lat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173840.1 | NP_776212.1 | 8 - 9 | 404 | 110 | |
| XM_006249160.2 | XP_006249222.1 | 9 - 10 | 575 | 110 | ||
| XM_008769138.1 | XP_008767360.1 | 8 - 9 | 857 | 110 | ||
| XM_008769139.1 | XP_008767361.1 | 7 - 8 | 821 | 110 | ||
| GenBank mRNA | AY170849.1 | - | 8 - 9 | 404 | 110 | |
| FQ231408.1 | - | 6 - 7 | 264 | 110 | ||
| FQ235223.1 | - | 6 - 7 | 264 | 110 | ||
Set Membership: |
|