Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598442_m1
          See other LDLR GE Assays ›
          Gene Symbol:
          Ldlr
          Gene Name:
          low density lipoprotein receptor
          Gene Aliases:
          LDLRA
          Chromosome Location:
          Chr.8: 22750425 - 22773305 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598442_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ldlr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175762.2 NP_786938.1 5 - 6 973 76
          GenBank mRNA X13722.1 - 5 - 6 973 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000239.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ldlr

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          lipid metabolic process
          lipid transport
          endocytosis
          receptor-mediated endocytosis
          aging
          cholesterol metabolic process
          response to hormone
          positive regulation of triglyceride biosynthetic process
          regulation of phosphatidylcholine catabolic process
          phospholipid transport
          intestinal cholesterol absorption
          cholesterol transport
          response to estradiol
          response to glucagon
          low-density lipoprotein particle clearance
          lipoprotein metabolic process
          lipoprotein catabolic process
          cholesterol homeostasis
          response to estrogen
          cholesterol import
          regulation of cholesterol homeostasis
          glycoprotein binding
          low-density lipoprotein receptor activity
          calcium ion binding
          protein binding
          low-density lipoprotein particle binding
          very-low-density lipoprotein particle receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041877 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041877 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P836880 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700936808 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700517843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4429 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26456 - -
          Agilent Rat cDNA Microarray (G4105A) 700936808 - -
          Agilent Rat cDNA Microarray (G4105A) 700517843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908525 - -
          Affymetrix Rat Gene 1.0 ST Array 10908533 - -
          Affymetrix Rat Gene 1.0 ST Array 10908524 - -
          Affymetrix Rat Gene 1.0 ST Array 10908540 - -
          Affymetrix Rat Gene 1.0 ST Array 10908536 - -
          Affymetrix Rat Gene 1.0 ST Array 10908532 - -
          Affymetrix Rat Gene 1.0 ST Array 10908529 - -
          Affymetrix Rat Gene 1.0 ST Array 10908530 - -
          Affymetrix Rat Gene 1.0 ST Array 10908522 - -
          Affymetrix Rat Gene 1.0 ST Array 10908526 - -
          Affymetrix Rat Gene 1.0 ST Array 10908535 - -
          Affymetrix Rat Gene 1.0 ST Array 10908539 - -
          Affymetrix Rat Gene 1.0 ST Array 10908531 - -
          Affymetrix Rat Gene 1.0 ST Array 10908528 - -
          Affymetrix Rat Gene 1.0 ST Array 10908534 - -
          Affymetrix Rat Gene 1.0 ST Array 10908527 - -
          Affymetrix Rat Gene 1.0 ST Array 10908537 - -
          Affymetrix Rat Gene 1.0 ST Array 10908542 - -
          Affymetrix Rat Gene 1.0 ST Array 10908523 - -
          Affymetrix Rat Gene 1.0 ST Array 10908538 - -
          Affymetrix Rat Gene 1.0 ST Array 10908541 - -
          Affymetrix Rat Gene 1.1 ST Array 10908521 - -
          Rat Genome 230 2.0 Array 1376089_PM_at - -
          Rat Genome 230 2.0 Array 1388218_at - -
          Rat Genome 230 2.0 Array 1376089_at - -
          Rat Genome 230 2.0 Array 1388218_PM_at - -
          Affymetrix HT Rat Focus Array 1388218_PM_at - -
          Affymetrix HT Rat Focus Array 1388218_at - -
          Affymetrix Rat Clariom S Assay PSR0800001469.rn.1 TC0800000239.rn.1 RTA TC
          Rat Expression Set 230 1388218_PM_at - -
          Rat Expression Set 230 1388218_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908521 - -
          Rat Genome U34 Set X13722_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21548897 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369382 - -
          RatRef-12 Expression BeadChip ILMN_51128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline