Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00665059_m1
          See other CBR1 GE Assays ›
          Gene Symbol:
          Cbr1
          Gene Name:
          carbonyl reductase 1
          Gene Aliases:
          Cbr
          Chromosome Location:
          Chr.11: 33845417 - 33847793 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00665059_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cbr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019170.2 NP_062043.1 445 77
          GenBank mRNA BC105893.1 - 497 7366019 77
          X84349.1 - 445 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cbr1

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovulation from ovarian follicle
          drug metabolic process
          peptidyl-lysine modification
          response to lipopolysaccharide
          response to testosterone
          response to gonadotropin
          vitamin K metabolic process
          phylloquinone catabolic process
          oxidation-reduction process
          carbonyl reductase (NADPH) activity
          oxidoreductase activity
          15-hydroxyprostaglandin dehydrogenase (NADP+) activity
          prostaglandin-E2 9-reductase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036352 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036352 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036352 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288156 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750324 - -
          Affymetrix Rat Gene 1.0 ST Array 10750313 - -
          Affymetrix Rat Gene 1.0 ST Array 10750322 - -
          Affymetrix Rat Gene 1.0 ST Array 10750311 - -
          Affymetrix Rat Gene 1.0 ST Array 10750323 - -
          Affymetrix Rat Gene 1.0 ST Array 10750312 - -
          Affymetrix Rat Gene 1.0 ST Array 10750321 - -
          Affymetrix Rat Gene 1.0 ST Array 10750314 - -
          Affymetrix Rat Gene 1.1 ST Array 10750320 - -
          Affymetrix Rat Gene 1.1 ST Array 10750310 - -
          Rat Genome 230 2.0 Array 1368037_PM_at - -
          Rat Genome 230 2.0 Array 1368037_at - -
          Affymetrix HT Rat Focus Array 1368037_at - -
          Affymetrix HT Rat Focus Array 1368037_PM_at - -
          Rat Expression Set 230 1368037_PM_at - -
          Rat Expression Set 230 1368037_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750320 - -
          Affymetrix Rat Gene 1.0 ST Array 10750310 - -
          Rat Genome U34 Set D89069_f_at - -
          Rat Genome U34 Set D89069_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367105 - -
          RatRef-12 Expression BeadChip ILMN_48764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline