Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00666184_m1
          See other TAOK2 GE Assays ›
          Gene Symbol:
          Taok2
          Gene Name:
          TAO kinase 2
          Gene Aliases:
          Tao2
          Chromosome Location:
          Chr.1: 198301789 - 198322264 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00666184_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taok2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022702.1 NP_073193.1 13 - 14 1617 64
          XM_006230329.2 XP_006230391.1 13 - 14 3058 64
          XM_006230330.2 XP_006230392.1 12 - 13 1479 64
          XM_006230331.2 XP_006230393.1 13 - 14 2346 64
          XM_006230332.2 XP_006230394.1 13 - 14 2329 64
          XM_006230334.2 XP_006230396.1 13 - 14 2331 64
          GenBank mRNA AB290408.1 - 12 - 13 1443 64
          AF140556.2 - 13 - 14 1617 64
          FQ223093.1 - 13 - 14 2254 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100008130.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taok2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          protein phosphorylation
          cellular response to DNA damage stimulus
          regulation of cell shape
          actin cytoskeleton organization
          positive regulation of stress-activated MAPK cascade
          positive regulation of JNK cascade
          focal adhesion assembly
          stress-activated MAPK cascade
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          ATP binding
          mitogen-activated protein kinase kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104885 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155721 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155721 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155721 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P564712 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702124146 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702452070 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700233687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9922 - -
          Agilent Rat cDNA Microarray (G4105A) 702124146 - -
          Agilent Rat cDNA Microarray (G4105A) 702452070 - -
          Agilent Rat cDNA Microarray (G4105A) 700233687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725869 - -
          Affymetrix Rat Gene 1.0 ST Array 10725861 - -
          Affymetrix Rat Gene 1.0 ST Array 10725859 - -
          Affymetrix Rat Gene 1.0 ST Array 10725872 - -
          Affymetrix Rat Gene 1.0 ST Array 10725867 - -
          Affymetrix Rat Gene 1.0 ST Array 10725871 - -
          Affymetrix Rat Gene 1.0 ST Array 10725864 - -
          Affymetrix Rat Gene 1.0 ST Array 10725868 - -
          Affymetrix Rat Gene 1.0 ST Array 10725860 - -
          Affymetrix Rat Gene 1.0 ST Array 10725873 - -
          Affymetrix Rat Gene 1.0 ST Array 10725863 - -
          Affymetrix Rat Gene 1.0 ST Array 10725862 - -
          Affymetrix Rat Gene 1.0 ST Array 10725866 - -
          Affymetrix Rat Gene 1.0 ST Array 10725874 - -
          Affymetrix Rat Gene 1.0 ST Array 10725855 - -
          Affymetrix Rat Gene 1.0 ST Array 10725875 - -
          Affymetrix Rat Gene 1.0 ST Array 10725857 - -
          Affymetrix Rat Gene 1.0 ST Array 10725870 - -
          Affymetrix Rat Gene 1.0 ST Array 10725856 - -
          Affymetrix Rat Gene 1.0 ST Array 10725865 - -
          Affymetrix Rat Gene 1.1 ST Array 10725854 - -
          Rat Genome 230 2.0 Array 1387140_PM_at - -
          Rat Genome 230 2.0 Array 1387140_at - -
          Affymetrix HT Rat Focus Array 1387140_PM_at - -
          Affymetrix HT Rat Focus Array 1387140_at - -
          Affymetrix Rat Clariom S Assay PSR0100066866.rn.1 TC0100008130.rn.1 RTA TC
          Rat Expression Set 230 1387140_PM_at - -
          Rat Expression Set 230 1387140_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725854 - -
          Rat Genome U34 Set rc_AA956196_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21065363 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360309 - -
          RatRef-12 Expression BeadChip ILMN_63425 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline