Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00678341_g1
          See other DUSP1 GE Assays ›
          Gene Symbol:
          Dusp1
          Gene Name:
          dual specificity phosphatase 1
          Gene Aliases:
          3CH134, CL100, MKP-1, Mkp1, Ptpn16
          Chromosome Location:
          Chr.10: 16970642 - 16973425 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00678341_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053769.3 NP_446221.2 3 - 4 870 63
          GenBank mRNA S81478.1 - 3 - 4 844 63
          U02553.1 - 3 - 4 856 63
          X84004.1 - 3 - 4 893 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000306.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dusp1

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          inactivation of MAPK activity
          endoderm formation
          protein dephosphorylation
          response to light stimulus
          response to organic substance
          response to estradiol
          response to retinoic acid
          cellular response to hormone stimulus
          response to testosterone
          peptidyl-tyrosine dephosphorylation
          intracellular signal transduction
          peptidyl-threonine dephosphorylation
          response to hydrogen peroxide
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          response to glucocorticoid
          negative regulation of meiotic cell cycle
          response to cAMP
          response to calcium ion
          negative regulation of ERK1 and ERK2 cascade
          mitotic cell cycle arrest
          regulation of mitotic cell cycle spindle assembly checkpoint
          negative regulation of DNA biosynthetic process
          protein tyrosine phosphatase activity
          protein tyrosine/serine/threonine phosphatase activity
          protein tyrosine/threonine phosphatase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12927 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12927 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12927 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12927 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700187524 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700502654 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17485 - -
          Agilent Rat cDNA Microarray (G4105A) 700502654 - -
          Agilent Rat cDNA Microarray (G4105A) 700187524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732654 - -
          Affymetrix Rat Gene 1.0 ST Array 10732656 - -
          Affymetrix Rat Gene 1.0 ST Array 10732653 - -
          Affymetrix Rat Gene 1.0 ST Array 10732655 - -
          Affymetrix Rat Gene 1.1 ST Array 10732652 - -
          Rat Genome 230 2.0 Array 1368146_PM_at - -
          Rat Genome 230 2.0 Array 1368147_at - -
          Rat Genome 230 2.0 Array 1368146_at - -
          Rat Genome 230 2.0 Array 1368147_PM_at - -
          Affymetrix HT Rat Focus Array 1368146_PM_at - -
          Affymetrix HT Rat Focus Array 1368146_at - -
          Affymetrix HT Rat Focus Array 1368147_PM_at - -
          Affymetrix HT Rat Focus Array 1368147_at - -
          Affymetrix Rat Clariom S Assay PSR1000003028.rn.1 TC1000000306.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003029.rn.1 TC1000000306.rn.1 RTA TC
          Rat Expression Set 230 1368146_at - -
          Rat Expression Set 230 1368146_PM_at - -
          Rat Expression Set 230 1368147_at - -
          Rat Expression Set 230 1368147_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732652 - -
          Rat Genome U34 Set S74351_s_at - -
          Rat Genome U34 Set U02553cds_s_at - -
          Rat Genome U34 Set S81478_s_at - -
          Rat Genome U34 Set rc_AA955855_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20814712 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367486 - -
          RatRef-12 Expression BeadChip ILMN_68233 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline