Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00688999_m1
          See other ARNT GE Assays ›
          Gene Symbol:
          Arnt
          Gene Name:
          aryl hydrocarbon receptor nuclear translocator
          Gene Aliases:
          Arnt1
          Chromosome Location:
          Chr.2: 196594175 - 196651937 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00688999_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arnt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012780.1 NP_036912.1 21 - 22 2320 94
          XM_006232850.2 XP_006232912.1 21 - 22 2480 94
          XM_006232852.2 XP_006232914.1 21 - 22 2570 94
          XM_006232853.2 XP_006232915.1 20 - 21 2435 94
          XM_008761280.1 XP_008759502.1 21 - 22 2477 94
          XM_008761281.1 XP_008759503.1 20 - 21 2432 94
          GenBank mRNA AY264361.1 - 21 - 22 2320 94
          AY264362.1 - 20 - 21 2275 94
          AY264363.1 - 21 - 22 2179 94
          AY264364.1 - 21 - 22 2305 94
          AY264365.1 - 21 - 22 2323 94
          U61184.1 - 21 - 22 2320 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001718.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arnt

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          embryonic placenta development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          signal transduction
          response to toxic substance
          positive regulation of vascular endothelial growth factor production
          cell differentiation
          intracellular receptor signaling pathway
          positive regulation of protein sumoylation
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hormone biosynthetic process
          regulation of nucleic acid-templated transcription
          transcription factor activity, transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          signal transducer activity
          aryl hydrocarbon receptor activity
          protein binding
          transcription factor binding
          aryl hydrocarbon receptor binding
          enhancer binding
          sequence-specific DNA binding
          protein heterodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11561 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11561 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11561 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11561 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701241377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64839 - -
          Agilent Rat cDNA Microarray (G4105A) 701241377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817405 - -
          Affymetrix Rat Gene 1.0 ST Array 10817410 - -
          Affymetrix Rat Gene 1.0 ST Array 10817398 - -
          Affymetrix Rat Gene 1.0 ST Array 10817403 - -
          Affymetrix Rat Gene 1.0 ST Array 10817411 - -
          Affymetrix Rat Gene 1.0 ST Array 10817412 - -
          Affymetrix Rat Gene 1.0 ST Array 10817413 - -
          Affymetrix Rat Gene 1.0 ST Array 10817417 - -
          Affymetrix Rat Gene 1.0 ST Array 10817407 - -
          Affymetrix Rat Gene 1.0 ST Array 10817397 - -
          Affymetrix Rat Gene 1.0 ST Array 10817402 - -
          Affymetrix Rat Gene 1.0 ST Array 10817414 - -
          Affymetrix Rat Gene 1.0 ST Array 10817404 - -
          Affymetrix Rat Gene 1.0 ST Array 10817409 - -
          Affymetrix Rat Gene 1.0 ST Array 10817408 - -
          Affymetrix Rat Gene 1.0 ST Array 10817415 - -
          Affymetrix Rat Gene 1.0 ST Array 10817399 - -
          Affymetrix Rat Gene 1.0 ST Array 10817400 - -
          Affymetrix Rat Gene 1.0 ST Array 10817406 - -
          Affymetrix Rat Gene 1.0 ST Array 10817416 - -
          Affymetrix Rat Gene 1.0 ST Array 10817418 - -
          Affymetrix Rat Gene 1.1 ST Array 10817396 - -
          Rat Genome 230 2.0 Array 1369244_PM_at - -
          Rat Genome 230 2.0 Array 1369244_at - -
          Affymetrix HT Rat Focus Array 1369244_at - -
          Affymetrix HT Rat Focus Array 1369244_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200011592.rn.1 TC0200001718.rn.1 RTA TC
          Rat Expression Set 230 1369244_at - -
          Rat Expression Set 230 1369244_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817396 - -
          Rat Genome U34 Set U61184_at - -
          Rat Genome U34 Set U61184_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20733981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353569 - -
          RatRef-12 Expression BeadChip ILMN_57269 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline