Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00691926_m1
          See other TIMM44 GE Assays ›
          Gene Symbol:
          Timm44
          Gene Name:
          translocase of inner mitochondrial membrane 44 homolog (yeast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 2517006 - 2533707 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00691926_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timm44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017267.1 NP_058963.1 11 - 12 1155 59
          GenBank mRNA AB006452.1 - 11 - 12 1155 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000048.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timm44

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          protein import into mitochondrial matrix
          protein transmembrane transport
          ATP binding
          P-P-bond-hydrolysis-driven protein transmembrane transporter activity
          enzyme binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P170396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170396 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P170396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P170396 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702025132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21869 - -
          Agilent Rat cDNA Microarray (G4105A) 702025132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756100 - -
          Affymetrix Rat Gene 1.0 ST Array 10756090 - -
          Affymetrix Rat Gene 1.0 ST Array 10756098 - -
          Affymetrix Rat Gene 1.0 ST Array 10756092 - -
          Affymetrix Rat Gene 1.0 ST Array 10756095 - -
          Affymetrix Rat Gene 1.0 ST Array 10756101 - -
          Affymetrix Rat Gene 1.0 ST Array 10756099 - -
          Affymetrix Rat Gene 1.0 ST Array 10756089 - -
          Affymetrix Rat Gene 1.0 ST Array 10756096 - -
          Affymetrix Rat Gene 1.0 ST Array 10756093 - -
          Affymetrix Rat Gene 1.0 ST Array 10756091 - -
          Affymetrix Rat Gene 1.0 ST Array 10756097 - -
          Affymetrix Rat Gene 1.0 ST Array 10756094 - -
          Affymetrix Rat Gene 1.1 ST Array 10756088 - -
          Rat Genome 230 2.0 Array 1398261_PM_at - -
          Rat Genome 230 2.0 Array 1391940_PM_at - -
          Rat Genome 230 2.0 Array 1391940_at - -
          Rat Genome 230 2.0 Array 1398261_at - -
          Affymetrix HT Rat Focus Array 1391940_PM_at - -
          Affymetrix HT Rat Focus Array 1391940_at - -
          Affymetrix HT Rat Focus Array 1398261_at - -
          Affymetrix HT Rat Focus Array 1398261_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200000495.rn.1 TC1200000048.rn.1 RTA TC
          Rat Expression Set 230 1398261_at - -
          Rat Expression Set 230 1398261_PM_at - -
          Rat Expression Set 230 1391940_at - -
          Rat Expression Set 230 1391940_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756088 - -
          Rat Genome U34 Set rc_AI045558_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22097140 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372640 - -
          RatRef-12 Expression BeadChip ILMN_63618 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline