Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710172_m1
          See other ABCA1 GE Assays ›
          Gene Symbol:
          Abca1
          Gene Name:
          ATP-binding cassette, subfamily A (ABC1), member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 69857717 - 69983036 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710172_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abca1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178095.2 NP_835196.1 2 - 3 239 76
          XM_008763708.1 XP_008761930.1 5 - 6 822 76
          XM_008763709.1 XP_008761931.1 5 - 6 822 76
          XM_008763710.1 XP_008761932.1 5 - 6 821 76
          GenBank mRNA AY208182.1 - 2 - 3 239 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002749.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abca1

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          peptide secretion
          protein lipidation
          phagocytosis, engulfment
          lysosome organization
          G-protein coupled receptor signaling pathway
          response to nutrient
          cholesterol metabolic process
          positive regulation of cholesterol efflux
          phospholipid transport
          endosomal transport
          cholesterol transport
          positive regulation of cAMP biosynthetic process
          intracellular cholesterol transport
          regulation of Cdc42 protein signal transduction
          cholesterol efflux
          phospholipid efflux
          ion transmembrane transport
          high-density lipoprotein particle assembly
          response to laminar fluid shear stress
          apolipoprotein A-I-mediated signaling pathway
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          response to drug
          cholesterol homeostasis
          reverse cholesterol transport
          phospholipid translocation
          interleukin-1 beta secretion
          transmembrane transport
          phospholipid homeostasis
          response to low-density lipoprotein particle
          response to high density lipoprotein particle
          platelet dense granule organization
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to cholesterol
          anion transmembrane transport
          receptor binding
          ATP binding
          phospholipid transporter activity
          anion transmembrane transporter activity
          cholesterol transporter activity
          syntaxin binding
          small GTPase binding
          apolipoprotein binding
          apolipoprotein A-I binding
          apolipoprotein A-I receptor activity
          ATPase activity, coupled to transmembrane movement of substances
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P323106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165187 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323106 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323106 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P323106 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P323106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43747 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10208 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52397 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24121 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68963 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45985 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876792 - -
          Affymetrix Rat Gene 1.0 ST Array 10876806 - -
          Affymetrix Rat Gene 1.0 ST Array 10876794 - -
          Affymetrix Rat Gene 1.0 ST Array 10876777 - -
          Affymetrix Rat Gene 1.0 ST Array 10876788 - -
          Affymetrix Rat Gene 1.0 ST Array 10876780 - -
          Affymetrix Rat Gene 1.0 ST Array 10876793 - -
          Affymetrix Rat Gene 1.0 ST Array 10876800 - -
          Affymetrix Rat Gene 1.0 ST Array 10876772 - -
          Affymetrix Rat Gene 1.0 ST Array 10876773 - -
          Affymetrix Rat Gene 1.0 ST Array 10876790 - -
          Affymetrix Rat Gene 1.0 ST Array 10876811 - -
          Affymetrix Rat Gene 1.0 ST Array 10876779 - -
          Affymetrix Rat Gene 1.0 ST Array 10876786 - -
          Affymetrix Rat Gene 1.0 ST Array 10876813 - -
          Affymetrix Rat Gene 1.0 ST Array 10876816 - -
          Affymetrix Rat Gene 1.0 ST Array 10876784 - -
          Affymetrix Rat Gene 1.0 ST Array 10876810 - -
          Affymetrix Rat Gene 1.0 ST Array 10876798 - -
          Affymetrix Rat Gene 1.0 ST Array 10876783 - -
          Affymetrix Rat Gene 1.0 ST Array 10876805 - -
          Affymetrix Rat Gene 1.0 ST Array 10876803 - -
          Affymetrix Rat Gene 1.0 ST Array 10876789 - -
          Affymetrix Rat Gene 1.0 ST Array 10876785 - -
          Affymetrix Rat Gene 1.0 ST Array 10876808 - -
          Affymetrix Rat Gene 1.0 ST Array 10876787 - -
          Affymetrix Rat Gene 1.0 ST Array 10876804 - -
          Affymetrix Rat Gene 1.0 ST Array 10876801 - -
          Affymetrix Rat Gene 1.0 ST Array 10876774 - -
          Affymetrix Rat Gene 1.0 ST Array 10876797 - -
          Affymetrix Rat Gene 1.0 ST Array 10876778 - -
          Affymetrix Rat Gene 1.0 ST Array 10876799 - -
          Affymetrix Rat Gene 1.0 ST Array 10876795 - -
          Affymetrix Rat Gene 1.0 ST Array 10876791 - -
          Affymetrix Rat Gene 1.0 ST Array 10876814 - -
          Affymetrix Rat Gene 1.0 ST Array 10876815 - -
          Affymetrix Rat Gene 1.0 ST Array 10876812 - -
          Affymetrix Rat Gene 1.0 ST Array 10876781 - -
          Affymetrix Rat Gene 1.0 ST Array 10876802 - -
          Affymetrix Rat Gene 1.0 ST Array 10876796 - -
          Affymetrix Rat Gene 1.0 ST Array 10876770 - -
          Affymetrix Rat Gene 1.0 ST Array 10876771 - -
          Affymetrix Rat Gene 1.0 ST Array 10876782 - -
          Affymetrix Rat Gene 1.1 ST Array 10876769 - -
          Rat Genome 230 2.0 Array 1383355_at - -
          Rat Genome 230 2.0 Array 1394490_PM_at - -
          Rat Genome 230 2.0 Array 1394490_at - -
          Rat Genome 230 2.0 Array 1383355_PM_at - -
          Rat Genome 230 2.0 Array 1384381_PM_at - -
          Rat Genome 230 2.0 Array 1384381_at - -
          Affymetrix HT Rat Focus Array 1383355_PM_at - -
          Affymetrix HT Rat Focus Array 1384381_PM_at - -
          Affymetrix HT Rat Focus Array 1383355_at - -
          Affymetrix HT Rat Focus Array 1384381_at - -
          Affymetrix Rat Clariom S Assay PSR0500020459.rn.1 TC0500002749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020460.rn.1 TC0500002749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020461.rn.1 TC0500002749.rn.1 RTA TC
          Rat Expression Set 230 1384381_at - -
          Rat Expression Set 230 1383355_at - -
          Rat Expression Set 230 1384381_PM_at - -
          Rat Expression Set 230 1394490_at - -
          Rat Expression Set 230 1383355_PM_at - -
          Rat Expression Set 230 1394490_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876769 - -
          Rat Genome U34 Set rc_AA964962_at - -
          Rat Genome U34 Set rc_AI171534_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21095557 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650701 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline