Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710383_m1
          See other DFNB31 GE Assays ›
          Gene Symbol:
          Dfnb31
          Gene Name:
          deafness, autosomal recessive 31
          Gene Aliases:
          Cip98, Whrn
          Chromosome Location:
          Chr.5: 79235541 - 79317206 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710383_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dfnb31
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181088.1 NP_851602.1 2 - 3 838 72
          GenBank mRNA AY227205.1 - 2 - 3 838 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002891.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dfnb31

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          sensory perception of sound
          positive regulation of gene expression
          sensory perception of light stimulus
          inner ear receptor stereocilium organization
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P165513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57809 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143054 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160667 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P165513 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P165513 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P165513 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P165513 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10593 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49553 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877517 - -
          Affymetrix Rat Gene 1.0 ST Array 10877522 - -
          Affymetrix Rat Gene 1.0 ST Array 10877511 - -
          Affymetrix Rat Gene 1.0 ST Array 10877514 - -
          Affymetrix Rat Gene 1.0 ST Array 10877523 - -
          Affymetrix Rat Gene 1.0 ST Array 10877519 - -
          Affymetrix Rat Gene 1.0 ST Array 10877508 - -
          Affymetrix Rat Gene 1.0 ST Array 10877509 - -
          Affymetrix Rat Gene 1.0 ST Array 10877515 - -
          Affymetrix Rat Gene 1.0 ST Array 10877520 - -
          Affymetrix Rat Gene 1.0 ST Array 10877512 - -
          Affymetrix Rat Gene 1.0 ST Array 10877516 - -
          Affymetrix Rat Gene 1.0 ST Array 10877510 - -
          Affymetrix Rat Gene 1.0 ST Array 10877521 - -
          Affymetrix Rat Gene 1.1 ST Array 10877507 - -
          Rat Genome 230 2.0 Array 1372434_PM_at - -
          Rat Genome 230 2.0 Array 1372434_at - -
          Affymetrix HT Rat Focus Array 1372434_at - -
          Affymetrix HT Rat Focus Array 1372434_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500021691.rn.1 TC0500002891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021693.rn.1 TC0500002891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021696.rn.1 TC0500002891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021692.rn.1 TC0500002891.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021695.rn.1 TC0500002891.rn.1 RTA TC
          Rat Expression Set 230 1372434_at - -
          Rat Expression Set 230 1372434_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877507 - -
          Rat Genome U34 Set rc_AI112639_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21257081 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363569 - -
          RatRef-12 Expression BeadChip ILMN_61567 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline