Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710679_m1
          See other OTOP1 GE Assays ›
          Gene Symbol:
          Otop1
          Gene Name:
          otopetrin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 77236278 - 77263729 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710679_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Otop1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181433.1 NP_852098.1 3 - 4 619 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002021.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Otop1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          detection of gravity
          biomineral tissue development
          inner ear morphogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P376940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P376940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P376940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P376940 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P376940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36543 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62180 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777314 - -
          Affymetrix Rat Gene 1.0 ST Array 10777315 - -
          Affymetrix Rat Gene 1.0 ST Array 10777319 - -
          Affymetrix Rat Gene 1.0 ST Array 10777318 - -
          Affymetrix Rat Gene 1.0 ST Array 10777317 - -
          Affymetrix Rat Gene 1.0 ST Array 10777320 - -
          Affymetrix Rat Gene 1.0 ST Array 10777316 - -
          Affymetrix Rat Gene 1.1 ST Array 10777313 - -
          Affymetrix Rat Clariom S Assay PSR1400014309.rn.1 TC1400002021.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014308.rn.1 TC1400002021.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014310.rn.1 TC1400002021.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10777313 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21405625 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358158 - -
          RatRef-12 Expression BeadChip ILMN_67544 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline