Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00711100_m1
          See other SYNM GE Assays ›
          Gene Symbol:
          Synm
          Gene Name:
          synemin, intermediate filament protein
          Gene Aliases:
          Dmn
          Chromosome Location:
          Chr.1: 128692112 - 128722048 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00711100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Synm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134858.3 NP_001128330.1 3 - 4 1239 128
          XM_006229337.2 XP_006229399.1 3 - 4 1234 128
          GenBank mRNA AB091769.1 - 3 - 4 1147 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005720.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          structural molecule activity
          structural constituent of cytoskeleton
          structural constituent of muscle
          vinculin binding
          intermediate filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P20719 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1870 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20719 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20719 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P20719 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10689 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10689 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10722698 - -
          Affymetrix Rat Gene 1.0 ST Array 10722700 - -
          Affymetrix Rat Gene 1.0 ST Array 10722697 - -
          Affymetrix Rat Gene 1.0 ST Array 10722705 - -
          Affymetrix Rat Gene 1.0 ST Array 10722699 - -
          Affymetrix Rat Gene 1.0 ST Array 10722696 - -
          Affymetrix Rat Gene 1.0 ST Array 10722704 - -
          Affymetrix Rat Gene 1.0 ST Array 10722701 - -
          Affymetrix Rat Gene 1.0 ST Array 10722695 - -
          Affymetrix Rat Gene 1.0 ST Array 10722703 - -
          Affymetrix Rat Gene 1.1 ST Array 10722694 - -
          Rat Genome 230 2.0 Array 1398113_at - -
          Rat Genome 230 2.0 Array 1376604_PM_at - -
          Rat Genome 230 2.0 Array 1398113_PM_at - -
          Rat Genome 230 2.0 Array 1372658_PM_at - -
          Rat Genome 230 2.0 Array 1372658_at - -
          Rat Genome 230 2.0 Array 1376604_at - -
          Affymetrix HT Rat Focus Array 1398113_PM_at - -
          Affymetrix HT Rat Focus Array 1372658_at - -
          Affymetrix HT Rat Focus Array 1398113_at - -
          Affymetrix HT Rat Focus Array 1372658_PM_at - -
          Affymetrix HT Rat Focus Array 1376604_PM_at - -
          Affymetrix HT Rat Focus Array 1376604_at - -
          Affymetrix Rat Clariom S Assay PSR0100045889.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045883.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045887.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045888.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045884.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045890.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045885.rn.1 TC0100005720.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045891.rn.1 TC0100005720.rn.1 RTA TC
          Rat Expression Set 230 1372658_PM_at - -
          Rat Expression Set 230 1376604_at - -
          Rat Expression Set 230 1372658_at - -
          Rat Expression Set 230 1376604_PM_at - -
          Rat Expression Set 230 1398113_PM_at - -
          Rat Expression Set 230 1398113_at - -
          Affymetrix Rat Gene 1.0 ST Array 10722694 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22108659 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373384 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline