Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755484_m1
          See other ALDH1A1 GE Assays ›
          Gene Symbol:
          Aldh1a1
          Gene Name:
          aldehyde dehydrogenase 1 family, member A1
          Gene Aliases:
          Ahd2, Aldh1, Aldh2
          Chromosome Location:
          Chr.1: 238222689 - 238264381 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755484_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022407.3 NP_071852.2 11 - 12 1386 77
          GenBank mRNA AF001896.1 - 11 - 12 1386 77
          AF001897.1 - 11 - 12 1386 77
          AF001898.1 - 11 - 12 1386 77
          BC061526.1 - 11 - 12 1405 6921926 77
          L42009.1 - 11 - 12 1382 77
          U79118.1 - 11 - 12 1385 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003337.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh1a1

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          liver development
          optic cup morphogenesis involved in camera-type eye development
          retinoic acid biosynthetic process
          response to oxidative stress
          midgut development
          response to organic cyclic compound
          response to estradiol
          response to retinoic acid
          response to drug
          retinol metabolic process
          retinoic acid metabolic process
          9-cis-retinoic acid biosynthetic process
          9-cis-retinoic acid metabolic process
          positive regulation of apoptotic process
          estrous cycle
          response to ethanol
          embryonic eye morphogenesis
          protein homotetramerization
          oxidation-reduction process
          retinal dehydrogenase activity
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD) activity
          benzaldehyde dehydrogenase (NAD+) activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71153 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067659 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067659 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365286 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700509013 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44515 - -
          Agilent Rat cDNA Microarray (G4105A) 700509013 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714337 - -
          Affymetrix Rat Gene 1.0 ST Array 10714333 - -
          Affymetrix Rat Gene 1.0 ST Array 10714326 - -
          Affymetrix Rat Gene 1.0 ST Array 10714329 - -
          Affymetrix Rat Gene 1.0 ST Array 10714328 - -
          Affymetrix Rat Gene 1.0 ST Array 10714330 - -
          Affymetrix Rat Gene 1.0 ST Array 10714327 - -
          Affymetrix Rat Gene 1.0 ST Array 10714332 - -
          Affymetrix Rat Gene 1.0 ST Array 10714334 - -
          Affymetrix Rat Gene 1.0 ST Array 10714336 - -
          Affymetrix Rat Gene 1.0 ST Array 10714335 - -
          Affymetrix Rat Gene 1.0 ST Array 10714325 - -
          Affymetrix Rat Gene 1.0 ST Array 10714331 - -
          Affymetrix Rat Gene 1.0 ST Array 10714324 - -
          Affymetrix Rat Gene 1.1 ST Array 10714323 - -
          Rat Genome 230 2.0 Array 1387022_PM_at - -
          Rat Genome 230 2.0 Array 1387022_at - -
          Affymetrix HT Rat Focus Array 1387022_PM_at - -
          Affymetrix HT Rat Focus Array 1387022_at - -
          Affymetrix Rat Clariom S Assay PSR0100027255.rn.1 TC0100003337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027256.rn.1 TC0100003337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027261.rn.1 TC0100003337.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027257.rn.1 TC0100003337.rn.1 RTA TC
          Rat Expression Set 230 1387022_PM_at - -
          Rat Expression Set 230 1387022_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714323 - -
          Rat Genome U34 Set AF001898_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20872072 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367475 - -
          RatRef-12 Expression BeadChip ILMN_48166 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline