Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00756297_m1
          See other UST4R GE Assays ›
          Gene Symbol:
          UST4r
          Gene Name:
          integral membrane transport protein UST4r
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 224319927 - 224389499 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00756297_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UST4r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134379.1 NP_599206.1 7 - 8 1401 125
          GenBank mRNA AJ132859.1 - 7 - 8 1401 125
          BC100065.1 - 1291 7373541 125
          FQ210050.1 - 1322 125
          FQ218143.1 - 1461 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007211.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UST4r

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73504 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134773 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13253 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13253 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13253 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84053 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11539 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728473 - -
          Affymetrix Rat Gene 1.0 ST Array 10728474 - -
          Affymetrix Rat Gene 1.0 ST Array 10728477 - -
          Affymetrix Rat Gene 1.0 ST Array 10728475 - -
          Affymetrix Rat Gene 1.0 ST Array 10728470 - -
          Affymetrix Rat Gene 1.0 ST Array 10728469 - -
          Affymetrix Rat Gene 1.0 ST Array 10728467 - -
          Affymetrix Rat Gene 1.0 ST Array 10728471 - -
          Affymetrix Rat Gene 1.0 ST Array 10728468 - -
          Affymetrix Rat Gene 1.0 ST Array 10728476 - -
          Affymetrix Rat Gene 1.0 ST Array 10728472 - -
          Affymetrix Rat Gene 1.1 ST Array 10728466 - -
          Rat Genome 230 2.0 Array 1369840_at - -
          Rat Genome 230 2.0 Array 1369840_PM_at - -
          Affymetrix HT Rat Focus Array 1369840_at - -
          Affymetrix HT Rat Focus Array 1369840_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100058113.rn.1 TC0100007211.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058109.rn.1 TC0100007211.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058108.rn.1 TC0100007211.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058112.rn.1 TC0100007211.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058115.rn.1 TC0100007211.rn.1 RTA TC
          Rat Expression Set 230 1369840_PM_at - -
          Rat Expression Set 230 1369840_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728466 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22397371 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350685 - -
          RatRef-12 Expression BeadChip ILMN_67034 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline