Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820586_g1
          See other AKR1B1 GE Assays ›
          Gene Symbol:
          Akr1b1
          Gene Name:
          aldo-keto reductase family 1, member B1 (aldose reductase)
          Gene Aliases:
          ALDRED, ALR-P-I, Akr1b3, Akr1b4, Aldr1, Alr, RATALDRED
          Chromosome Location:
          Chr.4: 61706866 - 61720959 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820586_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr1b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012498.1 NP_036630.1 4 - 5 462 69
          GenBank mRNA BC062034.1 - 4 - 5 447 5623700 69
          X05884.1 - 4 - 5 459 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr1b1

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          monosaccharide metabolic process
          sorbitol biosynthetic process
          response to water deprivation
          response to organic substance
          naphthalene metabolic process
          stress-activated protein kinase signaling cascade
          norepinephrine metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          positive regulation of JAK-STAT cascade
          positive regulation of smooth muscle cell proliferation
          oxidation-reduction process
          maternal process involved in female pregnancy
          cellular response to hydrogen peroxide
          inner medullary collecting duct development
          response to thyroid hormone
          cellular response to methylglyoxal
          cellular response to peptide
          alditol:NADP+ 1-oxidoreductase activity
          glyceraldehyde oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P784448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5930 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P784448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P784448 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P784448 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P432452 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700540855 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17276 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8402 - -
          Agilent Rat cDNA Microarray (G4105A) 700540855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864895 - -
          Affymetrix Rat Gene 1.0 ST Array 10861741 - -
          Affymetrix Rat Gene 1.0 ST Array 10861738 - -
          Affymetrix Rat Gene 1.0 ST Array 10861740 - -
          Affymetrix Rat Gene 1.0 ST Array 10861744 - -
          Affymetrix Rat Gene 1.0 ST Array 10861743 - -
          Affymetrix Rat Gene 1.0 ST Array 10861739 - -
          Affymetrix Rat Gene 1.0 ST Array 10861737 - -
          Affymetrix Rat Gene 1.0 ST Array 10861742 - -
          Affymetrix Rat Gene 1.1 ST Array 10864893 - -
          Affymetrix Rat Gene 1.1 ST Array 10861736 - -
          Rat Genome 230 2.0 Array 1367734_PM_at - -
          Rat Genome 230 2.0 Array 1367734_at - -
          Affymetrix HT Rat Focus Array 1396360_at - -
          Affymetrix HT Rat Focus Array 1396360_PM_at - -
          Affymetrix HT Rat Focus Array 1367734_at - -
          Affymetrix HT Rat Focus Array 1367734_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400018844.rn.1 TC0400002662.rn.1 RTA TC
          Rat Expression Set 230 1367734_PM_at - -
          Rat Expression Set 230 1367734_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864893 - -
          Affymetrix Rat Gene 1.0 ST Array 10861736 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22170446 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363873 - -
          RatRef-12 Expression BeadChip ILMN_64402 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline