Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821336_g1
          See other YBX1 GE Assays ›
          Gene Symbol:
          Ybx1
          Gene Name:
          Y box binding protein 1
          Gene Aliases:
          Byb1, Cbfa, Dbpb, Ef1a, Msy1, Nsep1, Yb1
          Chromosome Location:
          Chr.5: 138319753 - 138336721 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821336_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ybx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031563.3 NP_113751.3 2 - 3 344 96
          XM_008764058.1 XP_008762280.1 2 - 3 384 96
          XM_008764059.1 XP_008762281.1 2 - 3 584 96
          GenBank mRNA AB046928.2 - 2 - 3 503 96
          BC072486.1 - 2 - 3 344 7105037 96
          BC098672.1 - 2 - 3 344 7124218 96
          BC103637.1 - 2 - 3 338 7107271 96
          D13309.1 - 2 - 3 314 96
          M57299.1 - 2 - 3 320 96
          M69138.1 - 2 - 3 480 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ybx1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          positive regulation of cell proliferation
          RNA splicing
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin receptor signaling pathway
          negative regulation of striated muscle cell differentiation
          positive regulation of cell division
          CRD-mediated mRNA stabilization
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          p53 binding
          DNA binding
          chromatin binding
          single-stranded DNA binding
          RNA binding
          mRNA binding
          transcription factor binding
          sequence-specific DNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009810 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24111 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52119 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009809 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009809 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009810 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009809 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P109617 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55266 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33657 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10879377 - -
          Affymetrix Rat Gene 1.0 ST Array 10742743 - -
          Affymetrix Rat Gene 1.0 ST Array 10879373 - -
          Affymetrix Rat Gene 1.0 ST Array 10879376 - -
          Affymetrix Rat Gene 1.0 ST Array 10725157 - -
          Affymetrix Rat Gene 1.0 ST Array 10879374 - -
          Affymetrix Rat Gene 1.0 ST Array 10725158 - -
          Affymetrix Rat Gene 1.0 ST Array 10742742 - -
          Affymetrix Rat Gene 1.0 ST Array 10879378 - -
          Affymetrix Rat Gene 1.0 ST Array 10879379 - -
          Affymetrix Rat Gene 1.0 ST Array 10725156 - -
          Affymetrix Rat Gene 1.0 ST Array 10879375 - -
          Affymetrix Rat Gene 1.0 ST Array 10742741 - -
          Affymetrix Rat Gene 1.1 ST Array 10725155 - -
          Affymetrix Rat Gene 1.1 ST Array 10879372 - -
          Affymetrix Rat Gene 1.1 ST Array 10742740 - -
          Rat Genome 230 2.0 Array 1370213_at - -
          Rat Genome 230 2.0 Array 1370213_PM_at - -
          Affymetrix HT Rat Focus Array 1370213_PM_at - -
          Affymetrix HT Rat Focus Array 1370213_at - -
          Affymetrix Rat Clariom S Assay PSR0500025874.rn.1 TC0500003495.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500025876.rn.1 TC0500003495.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500025873.rn.1 TC0500003495.rn.1 RTA TC
          Rat Expression Set 230 1370213_PM_at - -
          Rat Expression Set 230 1370213_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879372 - -
          Affymetrix Rat Gene 1.0 ST Array 10742740 - -
          Affymetrix Rat Gene 1.0 ST Array 10725155 - -
          Rat Genome U34 Set rc_AI072435_at - -
          Rat Genome U34 Set D13309_s_at - -
          Rat Genome U34 Set S83025_s_at - -
          Rat Genome U34 Set rc_AI230572_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21579955 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376984 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365375 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline