Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821480_g1
          See other PRDX6 GE Assays ›
          Gene Symbol:
          Prdx6
          Gene Name:
          peroxiredoxin 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 79077567 - 79088113 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821480_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053576.2 NP_446028.1 3 - 4 423 77
          GenBank mRNA AF014009.1 - 3 - 4 423 77
          FQ218168.1 - 3 - 4 452 77
          Y17295.1 - 437 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx6

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          response to oxidative stress
          phospholipid catabolic process
          bleb assembly
          hydrogen peroxide catabolic process
          oxidation-reduction process
          peroxidase activity
          glutathione peroxidase activity
          phospholipase A2 activity
          protein binding
          peroxiredoxin activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1057763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174521 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1057763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1057763 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1057763 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1057763 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8114 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769219 - -
          Affymetrix Rat Gene 1.0 ST Array 10769223 - -
          Affymetrix Rat Gene 1.0 ST Array 10769222 - -
          Affymetrix Rat Gene 1.0 ST Array 10769221 - -
          Affymetrix Rat Gene 1.0 ST Array 10769220 - -
          Affymetrix Rat Gene 1.1 ST Array 10769218 - -
          Rat Genome 230 2.0 Array 1367969_at - -
          Rat Genome 230 2.0 Array 1367969_PM_at - -
          Affymetrix HT Rat Focus Array 1367969_PM_at - -
          Affymetrix HT Rat Focus Array 1367969_at - -
          Affymetrix Rat Clariom S Assay PSR1300012281.rn.1 TC1300001751.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012277.rn.1 TC1300001751.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012279.rn.1 TC1300001751.rn.1 RTA TC
          Rat Expression Set 230 1367969_at - -
          Rat Expression Set 230 1367969_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769218 - -
          Rat Genome U34 Set AF014009_at - -
          Rat Genome U34 Set Y17295cds_s_at - -
          Rat Genome U34 Set rc_AA892041_at - -
          Rat Genome U34 Set rc_AA892041_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21855346 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351805 - -
          RatRef-12 Expression BeadChip ILMN_62993 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline