Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01169349_m1
          See other USP7 GE Assays ›
          Gene Symbol:
          Usp7
          Gene Name:
          ubiquitin specific peptidase 7 (herpes virus-associated)
          Gene Aliases:
          Hausp
          Chromosome Location:
          Chr.10: 6930462 - 7019910 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01169349_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024790.1 NP_001019961.1 25 - 26 2723 146
          XM_006245756.2 XP_006245818.1 26 - 27 2935 146
          XM_006245757.2 XP_006245819.1 25 - 26 3772 146
          XM_006245758.2 XP_006245820.1 25 - 26 2865 146
          XM_006245759.1 XP_006245821.1 25 - 26 2816 146
          GenBank mRNA AY641530.1 - 25 - 26 2723 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000088.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Usp7

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription-coupled nucleotide-excision repair
          regulation of transcription, DNA-templated
          proteolysis
          ubiquitin-dependent protein catabolic process
          multicellular organism development
          maintenance of DNA methylation
          histone deubiquitination
          protein deubiquitination
          regulation of protein stability
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of sequence-specific DNA binding transcription factor activity
          regulation of proteasomal protein catabolic process
          p53 binding
          cysteine-type endopeptidase activity
          thiol-dependent ubiquitin-specific protease activity
          protein C-terminus binding
          transcription factor binding
          ubiquitin protein ligase binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4035 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076395 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076395 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076395 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P340448 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14683 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23774 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65250 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89066 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731573 - -
          Affymetrix Rat Gene 1.0 ST Array 10731579 - -
          Affymetrix Rat Gene 1.0 ST Array 10731578 - -
          Affymetrix Rat Gene 1.0 ST Array 10731589 - -
          Affymetrix Rat Gene 1.0 ST Array 10731567 - -
          Affymetrix Rat Gene 1.0 ST Array 10731575 - -
          Affymetrix Rat Gene 1.0 ST Array 10731562 - -
          Affymetrix Rat Gene 1.0 ST Array 10731566 - -
          Affymetrix Rat Gene 1.0 ST Array 10731571 - -
          Affymetrix Rat Gene 1.0 ST Array 10731590 - -
          Affymetrix Rat Gene 1.0 ST Array 10731569 - -
          Affymetrix Rat Gene 1.0 ST Array 10731561 - -
          Affymetrix Rat Gene 1.0 ST Array 10731586 - -
          Affymetrix Rat Gene 1.0 ST Array 10731582 - -
          Affymetrix Rat Gene 1.0 ST Array 10731581 - -
          Affymetrix Rat Gene 1.0 ST Array 10731563 - -
          Affymetrix Rat Gene 1.0 ST Array 10731568 - -
          Affymetrix Rat Gene 1.0 ST Array 10731572 - -
          Affymetrix Rat Gene 1.0 ST Array 10731564 - -
          Affymetrix Rat Gene 1.0 ST Array 10731565 - -
          Affymetrix Rat Gene 1.0 ST Array 10731559 - -
          Affymetrix Rat Gene 1.0 ST Array 10731576 - -
          Affymetrix Rat Gene 1.0 ST Array 10731580 - -
          Affymetrix Rat Gene 1.0 ST Array 10731584 - -
          Affymetrix Rat Gene 1.0 ST Array 10731583 - -
          Affymetrix Rat Gene 1.0 ST Array 10731570 - -
          Affymetrix Rat Gene 1.0 ST Array 10731588 - -
          Affymetrix Rat Gene 1.0 ST Array 10731574 - -
          Affymetrix Rat Gene 1.0 ST Array 10731587 - -
          Affymetrix Rat Gene 1.0 ST Array 10731577 - -
          Affymetrix Rat Gene 1.0 ST Array 10731585 - -
          Affymetrix Rat Gene 1.1 ST Array 10731557 - -
          Rat Genome 230 2.0 Array 1398351_at - -
          Rat Genome 230 2.0 Array 1394393_at - -
          Rat Genome 230 2.0 Array 1385527_at - -
          Rat Genome 230 2.0 Array 1394393_PM_at - -
          Rat Genome 230 2.0 Array 1385527_PM_at - -
          Rat Genome 230 2.0 Array 1393042_at - -
          Rat Genome 230 2.0 Array 1398351_PM_at - -
          Rat Genome 230 2.0 Array 1393042_PM_at - -
          Affymetrix HT Rat Focus Array 1393042_at - -
          Affymetrix HT Rat Focus Array 1385527_PM_at - -
          Affymetrix HT Rat Focus Array 1385527_at - -
          Affymetrix HT Rat Focus Array 1394393_at - -
          Affymetrix HT Rat Focus Array 1393042_PM_at - -
          Affymetrix HT Rat Focus Array 1398351_PM_at - -
          Affymetrix HT Rat Focus Array 1394393_PM_at - -
          Affymetrix HT Rat Focus Array 1398351_at - -
          Affymetrix Rat Clariom S Assay PSR1000000846.rn.1 TC1000000088.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000000845.rn.1 TC1000000088.rn.1 RTA TC
          Rat Expression Set 230 1398351_PM_at - -
          Rat Expression Set 230 1398351_at - -
          Rat Expression Set 230 1393042_at - -
          Rat Expression Set 230 1385527_at - -
          Rat Expression Set 230 1393042_PM_at - -
          Rat Expression Set 230 1394393_at - -
          Rat Expression Set 230 1394393_PM_at - -
          Rat Expression Set 230 1385527_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731557 - -
          Rat Genome U34 Set rc_AI175593_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21188553 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369921 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline