Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01246634_m1
          See other CTNNBL1 GE Assays ›
          Gene Symbol:
          Ctnnbl1
          Gene Name:
          catenin, beta like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 154212245 - 154373170 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01246634_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctnnbl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024870.1 NP_001020041.1 15 - 16 1655 67
          GenBank mRNA BC097352.1 - 15 - 16 1655 7387542 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002179.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctnnbl1

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          apoptotic process
          biological_process
          RNA splicing
          somatic diversification of immunoglobulins
          positive regulation of apoptotic process
          molecular_function
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P625546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89664 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102685 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P625546 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P625546 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P625546 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21557 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P625546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26059 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58111 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85303 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101777 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54139 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33683 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841647 - -
          Affymetrix Rat Gene 1.0 ST Array 10841654 - -
          Affymetrix Rat Gene 1.0 ST Array 10841658 - -
          Affymetrix Rat Gene 1.0 ST Array 10841648 - -
          Affymetrix Rat Gene 1.0 ST Array 10841650 - -
          Affymetrix Rat Gene 1.0 ST Array 10841652 - -
          Affymetrix Rat Gene 1.0 ST Array 10841643 - -
          Affymetrix Rat Gene 1.0 ST Array 10841659 - -
          Affymetrix Rat Gene 1.0 ST Array 10841651 - -
          Affymetrix Rat Gene 1.0 ST Array 10841657 - -
          Affymetrix Rat Gene 1.0 ST Array 10841649 - -
          Affymetrix Rat Gene 1.0 ST Array 10841645 - -
          Affymetrix Rat Gene 1.0 ST Array 10841655 - -
          Affymetrix Rat Gene 1.0 ST Array 10841653 - -
          Affymetrix Rat Gene 1.0 ST Array 10841644 - -
          Affymetrix Rat Gene 1.0 ST Array 10841656 - -
          Affymetrix Rat Gene 1.0 ST Array 10841646 - -
          Affymetrix Rat Gene 1.1 ST Array 10841642 - -
          Rat Genome 230 2.0 Array 1371633_PM_at - -
          Rat Genome 230 2.0 Array 1371633_at - -
          Affymetrix HT Rat Focus Array 1371633_PM_at - -
          Affymetrix HT Rat Focus Array 1371633_at - -
          Affymetrix Rat Clariom S Assay PSR0300016154.rn.1 TC0300002179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300016155.rn.1 TC0300002179.rn.1 RTA TC
          Rat Expression Set 230 1371633_at - -
          Rat Expression Set 230 1371633_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841642 - -
          Rat Genome U34 Set rc_AI058714_s_at - -
          Rat Genome U34 Set rc_AI059023_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20790632 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline