Search
Search

Atxn7l3| Gene Symbol: Atxn7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247470.2 | XP_006247532.1 | 1 - 2 | 1363 | 64 | |
| XM_006247472.2 | XP_006247534.1 | 1 - 2 | 1362 | 64 | ||
| XM_008768223.1 | XP_008766445.1 | 1 - 2 | 1365 | 64 | ||
| XM_008768224.1 | XP_008766446.1 | 2 - 3 | 709 | 64 | ||
| XM_008768225.1 | XP_008766447.1 | 1 - 2 | 1364 | 64 | ||
Set Membership: |
|