Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400691_m1
          See other ATG9A GE Assays ›
          Gene Symbol:
          Atg9a
          Gene Name:
          autophagy related 9A
          Gene Aliases:
          RGD1310450
          Chromosome Location:
          Chr.9: 82382800 - 82393428 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01400691_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atg9a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014218.1 NP_001014240.1 13 - 14 2676 92
          XM_006245247.2 XP_006245309.1 13 - 14 2380 92
          XM_008767262.1 XP_008765484.1 13 - 14 3043 92
          XM_008767263.1 XP_008765485.1 13 - 14 2555 92
          XM_008767264.1 XP_008765486.1 13 - 14 2383 92
          XM_008767265.1 XP_008765487.1 14 - 15 2441 92
          XM_008767266.1 XP_008765488.1 14 - 15 2444 92
          GenBank mRNA BC089204.1 - 13 - 14 2676 7190326 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002262.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atg9a

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          autophagy
          protein transport
          protein localization to Golgi apparatus
          protein localization to pre-autophagosomal structure
          piecemeal microautophagy of nucleus
          late nucleophagy
          innate immune response
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157929 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142314 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157929 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157929 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157929 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157934 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P577938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929090 - -
          Affymetrix Rat Gene 1.0 ST Array 10929088 - -
          Affymetrix Rat Gene 1.0 ST Array 10929102 - -
          Affymetrix Rat Gene 1.0 ST Array 10929087 - -
          Affymetrix Rat Gene 1.0 ST Array 10929099 - -
          Affymetrix Rat Gene 1.0 ST Array 10929092 - -
          Affymetrix Rat Gene 1.0 ST Array 10929097 - -
          Affymetrix Rat Gene 1.0 ST Array 10929104 - -
          Affymetrix Rat Gene 1.0 ST Array 10929094 - -
          Affymetrix Rat Gene 1.0 ST Array 10929098 - -
          Affymetrix Rat Gene 1.0 ST Array 10929101 - -
          Affymetrix Rat Gene 1.0 ST Array 10929103 - -
          Affymetrix Rat Gene 1.0 ST Array 10929093 - -
          Affymetrix Rat Gene 1.0 ST Array 10929100 - -
          Affymetrix Rat Gene 1.0 ST Array 10929095 - -
          Affymetrix Rat Gene 1.0 ST Array 10929091 - -
          Affymetrix Rat Gene 1.0 ST Array 10929089 - -
          Affymetrix Rat Gene 1.1 ST Array 10929086 - -
          Rat Genome 230 2.0 Array 1396842_PM_at - -
          Rat Genome 230 2.0 Array 1396842_at - -
          Rat Genome 230 2.0 Array 1382807_PM_at - -
          Rat Genome 230 2.0 Array 1382807_at - -
          Affymetrix HT Rat Focus Array 1382807_PM_at - -
          Affymetrix HT Rat Focus Array 1382807_at - -
          Affymetrix HT Rat Focus Array 1396842_PM_at - -
          Affymetrix HT Rat Focus Array 1396842_at - -
          Affymetrix Rat Clariom S Assay PSR0900018483.rn.1 TC0900002262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900018481.rn.1 TC0900002262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900018482.rn.1 TC0900002262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900018480.rn.1 TC0900002262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900018479.rn.1 TC0900002262.rn.1 RTA TC
          Rat Expression Set 230 1396842_at - -
          Rat Expression Set 230 1382807_PM_at - -
          Rat Expression Set 230 1382807_at - -
          Rat Expression Set 230 1396842_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929086 - -
          Rat Genome U34 Set H32584_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21281597 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368406 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline