Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408819_m1
          See other CCDC93 GE Assays ›
          Gene Symbol:
          Ccdc93
          Gene Name:
          coiled-coil domain containing 93
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 37400460 - 37473973 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408819_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024997.1 NP_001020168.1 22 - 23 1856 72
          XM_008769456.1 XP_008767678.1 23 - 24 1890 72
          GenBank mRNA BC091337.1 - 22 - 23 1856 7314934 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000156.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc93

          Gene Ontology Categories:

          Function(s)

          Golgi to plasma membrane transport
          protein transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23729 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060151 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060151 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060151 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1001914 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25753 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45413 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90057 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52461 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57327 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85288 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763561 - -
          Affymetrix Rat Gene 1.0 ST Array 10763572 - -
          Affymetrix Rat Gene 1.0 ST Array 10763559 - -
          Affymetrix Rat Gene 1.0 ST Array 10763568 - -
          Affymetrix Rat Gene 1.0 ST Array 10763575 - -
          Affymetrix Rat Gene 1.0 ST Array 10763556 - -
          Affymetrix Rat Gene 1.0 ST Array 10763554 - -
          Affymetrix Rat Gene 1.0 ST Array 10763576 - -
          Affymetrix Rat Gene 1.0 ST Array 10763570 - -
          Affymetrix Rat Gene 1.0 ST Array 10763555 - -
          Affymetrix Rat Gene 1.0 ST Array 10763563 - -
          Affymetrix Rat Gene 1.0 ST Array 10763571 - -
          Affymetrix Rat Gene 1.0 ST Array 10763558 - -
          Affymetrix Rat Gene 1.0 ST Array 10763569 - -
          Affymetrix Rat Gene 1.0 ST Array 10763553 - -
          Affymetrix Rat Gene 1.0 ST Array 10763574 - -
          Affymetrix Rat Gene 1.0 ST Array 10763565 - -
          Affymetrix Rat Gene 1.0 ST Array 10763573 - -
          Affymetrix Rat Gene 1.0 ST Array 10763577 - -
          Affymetrix Rat Gene 1.0 ST Array 10763562 - -
          Affymetrix Rat Gene 1.0 ST Array 10763564 - -
          Affymetrix Rat Gene 1.0 ST Array 10763566 - -
          Affymetrix Rat Gene 1.0 ST Array 10763560 - -
          Affymetrix Rat Gene 1.0 ST Array 10763567 - -
          Affymetrix Rat Gene 1.1 ST Array 10763552 - -
          Rat Genome 230 2.0 Array 1385268_PM_at - -
          Rat Genome 230 2.0 Array 1385269_PM_s_at - -
          Rat Genome 230 2.0 Array 1385268_at - -
          Rat Genome 230 2.0 Array 1385269_s_at - -
          Affymetrix HT Rat Focus Array 1385268_at - -
          Affymetrix HT Rat Focus Array 1385269_s_at - -
          Affymetrix HT Rat Focus Array 1385269_PM_s_at - -
          Affymetrix HT Rat Focus Array 1385268_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300000811.rn.1 TC1300000156.rn.1 RTA TC
          Rat Expression Set 230 1385269_s_at - -
          Rat Expression Set 230 1385269_PM_s_at - -
          Rat Expression Set 230 1385268_at - -
          Rat Expression Set 230 1385268_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10763552 - -
          Rat Genome U34 Set rc_AA955847_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21408753 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373527 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline