Search
Search
| Gene Symbol: Arl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022700.2 | NP_073191.1 | 3 - 4 | 325 | 88 | |
| XM_006231559.2 | XP_006231621.1 | 4 - 5 | 846 | 88 | ||
| GenBank mRNA | BC084722.1 | - | 3 - 4 | 325 | 7310623 | 88 |
| U12568.1 | - | 3 - 4 | 329 | 88 | ||
| X76921.1 | - | 4 - 5 | 457 | 88 | ||
Set Membership: |
|