Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410988_g1
          See other SMNDC1 GE Assays ›
          Gene Symbol:
          Smndc1
          Gene Name:
          survival motor neuron domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 274096068 - 274107138 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410988_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smndc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025400.3 NP_001020571.1 5 - 6 605 96
          NM_001035234.2 NP_001030311.1 5 - 6 767 96
          NR_051995.1 5 - 6 784 96
          XM_006231608.2 XP_006231670.1 5 - 6 1044 96
          GenBank mRNA BC097986.1 - 5 - 6 605 7444662 96
          BC098697.1 - 5 - 6 746 7386640 96
          FQ220088.1 - 4 - 5 548 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smndc1

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal complex assembly
          apoptotic process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P667238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47444 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667238 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667238 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P667238 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P667238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730885 - -
          Affymetrix Rat Gene 1.0 ST Array 10739205 - -
          Affymetrix Rat Gene 1.0 ST Array 10730892 - -
          Affymetrix Rat Gene 1.0 ST Array 10730891 - -
          Affymetrix Rat Gene 1.0 ST Array 10730888 - -
          Affymetrix Rat Gene 1.0 ST Array 10730886 - -
          Affymetrix Rat Gene 1.0 ST Array 10730890 - -
          Affymetrix Rat Gene 1.0 ST Array 10730889 - -
          Affymetrix Rat Gene 1.1 ST Array 10739204 - -
          Affymetrix Rat Gene 1.1 ST Array 10730884 - -
          Rat Genome 230 2.0 Array 1382165_at - -
          Rat Genome 230 2.0 Array 1388979_PM_at - -
          Rat Genome 230 2.0 Array 1382165_PM_at - -
          Rat Genome 230 2.0 Array 1388979_at - -
          Affymetrix HT Rat Focus Array 1388979_PM_at - -
          Affymetrix HT Rat Focus Array 1382165_at - -
          Affymetrix HT Rat Focus Array 1382165_PM_at - -
          Affymetrix HT Rat Focus Array 1388979_at - -
          Affymetrix Rat Clariom S Assay PSR0100063742.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063741.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063743.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063736.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063745.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063737.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063744.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063746.rn.1 TC0100007904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100063748.rn.1 TC0100007904.rn.1 RTA TC
          Rat Expression Set 230 1388979_PM_at - -
          Rat Expression Set 230 1388979_at - -
          Rat Expression Set 230 1382165_PM_at - -
          Rat Expression Set 230 1382165_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739204 - -
          Affymetrix Rat Gene 1.0 ST Array 10730884 - -
          Rat Genome U34 Set rc_AA925359_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline