Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413743_m1
          See other BTRC GE Assays ›
          Gene Symbol:
          Btrc
          Gene Name:
          beta-transducin repeat containing E3 ubiquitin protein ligase
          Gene Aliases:
          beta-TrCP1
          Chromosome Location:
          Chr.1: 265100100 - 265270263 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413743_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Btrc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007148.2 NP_001007149.2 14 - 15 1791 72
          XM_006231529.2 XP_006231591.1 14 - 15 2374 72
          XM_006231530.2 XP_006231592.1 13 - 14 1785 72
          XM_006231531.2 XP_006231593.1 13 - 14 1751 72
          XM_006231535.2 XP_006231597.1 13 - 14 1636 72
          XM_006231536.2 XP_006231598.1 13 - 14 1718 72
          XM_006231537.2 XP_006231599.1 13 - 14 2265 72
          XM_006231538.2 XP_006231600.1 12 - 13 1675 72
          XM_006231539.2 XP_006231601.1 12 - 13 2421 72
          XM_006231541.2 XP_006231603.1 12 - 13 2135 72
          XM_006231543.2 XP_006231605.1 11 - 12 2343 72
          GenBank mRNA BC085125.1 - 13 - 14 1612 7131883 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003708.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Btrc

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          protein dephosphorylation
          ubiquitin-dependent protein catabolic process
          protein ubiquitination
          protein catabolic process
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          protein destabilization
          mammary gland epithelial cell proliferation
          regulation of circadian rhythm
          positive regulation of circadian rhythm
          regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          regulation of cell cycle
          branching involved in mammary gland duct morphogenesis
          regulation of proteasomal protein catabolic process
          cellular response to organic cyclic compound
          ubiquitin-protein transferase activity
          beta-catenin binding
          protein phosphorylated amino acid binding
          protein dimerization activity
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171146 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063052 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063052 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063052 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18668 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82994 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12158 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80635 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85169 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25932 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33653 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715690 - -
          Affymetrix Rat Gene 1.0 ST Array 10715681 - -
          Affymetrix Rat Gene 1.0 ST Array 10715689 - -
          Affymetrix Rat Gene 1.0 ST Array 10715692 - -
          Affymetrix Rat Gene 1.0 ST Array 10715693 - -
          Affymetrix Rat Gene 1.0 ST Array 10715695 - -
          Affymetrix Rat Gene 1.0 ST Array 10715687 - -
          Affymetrix Rat Gene 1.0 ST Array 10715682 - -
          Affymetrix Rat Gene 1.0 ST Array 10715684 - -
          Affymetrix Rat Gene 1.0 ST Array 10715680 - -
          Affymetrix Rat Gene 1.0 ST Array 10715686 - -
          Affymetrix Rat Gene 1.0 ST Array 10715696 - -
          Affymetrix Rat Gene 1.0 ST Array 10715694 - -
          Affymetrix Rat Gene 1.0 ST Array 10715691 - -
          Affymetrix Rat Gene 1.0 ST Array 10715685 - -
          Affymetrix Rat Gene 1.0 ST Array 10715683 - -
          Affymetrix Rat Gene 1.0 ST Array 10715688 - -
          Affymetrix Rat Gene 1.1 ST Array 10715679 - -
          Rat Genome 230 2.0 Array 1374654_at - -
          Rat Genome 230 2.0 Array 1374654_PM_at - -
          Affymetrix HT Rat Focus Array 1374654_PM_at - -
          Affymetrix HT Rat Focus Array 1374654_at - -
          Affymetrix Rat Clariom S Assay PSR0100030210.rn.1 TC0100003708.rn.1 RTA TC
          Rat Expression Set 230 1374654_PM_at - -
          Rat Expression Set 230 1374654_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715679 - -
          Rat Genome U34 Set rc_AI179552_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21679815 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352972 - -
          RatRef-12 Expression BeadChip ILMN_65477 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline