Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418502_m1
          See other ZW10 GE Assays ›
          Gene Symbol:
          Zw10
          Gene Name:
          zw10 kinetochore protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 53365881 - 53390742 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418502_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zw10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024801.1 NP_001019972.1 12 - 13 1832 79
          GenBank mRNA BC097452.1 - 12 - 13 1832 7456845 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000661.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zw10

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s)

          mitotic sister chromatid segregation
          establishment of mitotic spindle orientation
          protein complex assembly
          ER to Golgi vesicle-mediated transport
          Golgi organization
          mitotic metaphase plate congression
          mitotic cell cycle checkpoint
          regulation of exit from mitosis
          protein transport
          protein localization to kinetochore
          cell division

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P037358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137731 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037358 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037358 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P037358 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P481086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65610 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909820 - -
          Affymetrix Rat Gene 1.0 ST Array 10909826 - -
          Affymetrix Rat Gene 1.0 ST Array 10909814 - -
          Affymetrix Rat Gene 1.0 ST Array 10909815 - -
          Affymetrix Rat Gene 1.0 ST Array 10909821 - -
          Affymetrix Rat Gene 1.0 ST Array 10909822 - -
          Affymetrix Rat Gene 1.0 ST Array 10909817 - -
          Affymetrix Rat Gene 1.0 ST Array 10909819 - -
          Affymetrix Rat Gene 1.0 ST Array 10909825 - -
          Affymetrix Rat Gene 1.0 ST Array 10909823 - -
          Affymetrix Rat Gene 1.0 ST Array 10909827 - -
          Affymetrix Rat Gene 1.0 ST Array 10909818 - -
          Affymetrix Rat Gene 1.0 ST Array 10909813 - -
          Affymetrix Rat Gene 1.0 ST Array 10909816 - -
          Affymetrix Rat Gene 1.0 ST Array 10909812 - -
          Affymetrix Rat Gene 1.0 ST Array 10909824 - -
          Affymetrix Rat Gene 1.1 ST Array 10909811 - -
          Rat Genome 230 2.0 Array 1374938_at - -
          Rat Genome 230 2.0 Array 1374938_PM_at - -
          Affymetrix HT Rat Focus Array 1374938_at - -
          Affymetrix HT Rat Focus Array 1374938_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800004298.rn.1 TC0800000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004301.rn.1 TC0800000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004299.rn.1 TC0800000661.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004300.rn.1 TC0800000661.rn.1 RTA TC
          Rat Expression Set 230 1374938_PM_at - -
          Rat Expression Set 230 1374938_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909811 - -
          Rat Genome U34 Set rc_AI230413_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22223871 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline