Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423825_m1
          See other PARD6B GE Assays ›
          Gene Symbol:
          Pard6b
          Gene Name:
          par-6 family cell polarity regulator beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 164822111 - 164843395 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423825_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pard6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108609.1 NP_001102079.1 1 - 2 306 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002384.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s)

          protein complex assembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P537971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7950 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21006 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P537971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P537971 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P537971 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20914 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842492 - -
          Affymetrix Rat Gene 1.0 ST Array 10842489 - -
          Affymetrix Rat Gene 1.0 ST Array 10842488 - -
          Affymetrix Rat Gene 1.0 ST Array 10842490 - -
          Affymetrix Rat Gene 1.0 ST Array 10842491 - -
          Affymetrix Rat Gene 1.1 ST Array 10842486 - -
          Rat Genome 230 2.0 Array 1383224_at - -
          Rat Genome 230 2.0 Array 1383224_PM_at - -
          Affymetrix HT Rat Focus Array 1383224_PM_at - -
          Affymetrix HT Rat Focus Array 1383224_at - -
          Affymetrix Rat Clariom S Assay PSR0300017824.rn.1 TC0300002384.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017823.rn.1 TC0300002384.rn.1 RTA TC
          Rat Expression Set 230 1383224_at - -
          Rat Expression Set 230 1383224_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842486 - -
          Rat Genome U34 Set rc_AI059678_at - -
          Rat Genome U34 Set rc_AI178666_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21086206 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-89x45:80/100.66.77.245:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0