Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01429297_g1
          See other GLO1 GE Assays ›
          Gene Symbol:
          Glo1
          Gene Name:
          glyoxalase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 9273589 - 9291608 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01429297_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Glo1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207594.2 NP_997477.1 5 - 6 539 84
          GenBank mRNA BC061570.1 - 5 - 6 543 6918143 84
          FQ220772.1 - 5 - 6 538 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Glo1

          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          regulation of transcription from RNA polymerase II promoter
          glutathione metabolic process
          methylglyoxal metabolic process
          osteoclast differentiation
          negative regulation of apoptotic process
          lactoylglutathione lyase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P353553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P353553 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P353553 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P353553 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P353553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86256 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832121 - -
          Affymetrix Rat Gene 1.0 ST Array 10832118 - -
          Affymetrix Rat Gene 1.0 ST Array 10832116 - -
          Affymetrix Rat Gene 1.0 ST Array 10832119 - -
          Affymetrix Rat Gene 1.0 ST Array 10832120 - -
          Affymetrix Rat Gene 1.0 ST Array 10832117 - -
          Affymetrix Rat Gene 1.1 ST Array 10832115 - -
          Rat Genome 230 2.0 Array 1399098_at - -
          Rat Genome 230 2.0 Array 1399098_PM_at - -
          Affymetrix HT Rat Focus Array 1399098_PM_at - -
          Affymetrix HT Rat Focus Array 1399098_at - -
          Rat Expression Set 230 1399098_at - -
          Rat Expression Set 230 1399098_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10832115 - -
          Rat Genome U34 Set rc_AI229104_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21752333 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355977 - -
          RatRef-12 Expression BeadChip ILMN_58807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline