Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01432075_g1
          See other EDC4 GE Assays ›
          Gene Symbol:
          Edc4
          Gene Name:
          enhancer of mRNA decapping 4
          Gene Aliases:
          LRRG00115, Nrn1l, RGD1309136
          Chromosome Location:
          Chr.19: 37852659 - 37864651 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01432075_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Edc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033068.3 NP_001028240.3 27 - 28 4037 86
          GenBank mRNA BC103628.1 - 27 - 28 4037 7939290 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000496.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Edc4

          Panther Classification:

          Molecular Function -

          mRNA capping factor

          Gene Ontology Categories:

          Function(s)

          nervous system development

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P090819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090819 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090819 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P090819 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P624251 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405533 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P112335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64820 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807420 - -
          Affymetrix Rat Gene 1.0 ST Array 10807417 - -
          Affymetrix Rat Gene 1.0 ST Array 10807414 - -
          Affymetrix Rat Gene 1.0 ST Array 10807403 - -
          Affymetrix Rat Gene 1.0 ST Array 10807421 - -
          Affymetrix Rat Gene 1.0 ST Array 10807410 - -
          Affymetrix Rat Gene 1.0 ST Array 10807405 - -
          Affymetrix Rat Gene 1.0 ST Array 10807401 - -
          Affymetrix Rat Gene 1.0 ST Array 10807419 - -
          Affymetrix Rat Gene 1.0 ST Array 10807396 - -
          Affymetrix Rat Gene 1.0 ST Array 10807397 - -
          Affymetrix Rat Gene 1.0 ST Array 10807400 - -
          Affymetrix Rat Gene 1.0 ST Array 10807413 - -
          Affymetrix Rat Gene 1.0 ST Array 10807404 - -
          Affymetrix Rat Gene 1.0 ST Array 10807425 - -
          Affymetrix Rat Gene 1.0 ST Array 10807423 - -
          Affymetrix Rat Gene 1.0 ST Array 10807394 - -
          Affymetrix Rat Gene 1.0 ST Array 10807422 - -
          Affymetrix Rat Gene 1.0 ST Array 10807416 - -
          Affymetrix Rat Gene 1.0 ST Array 10807418 - -
          Affymetrix Rat Gene 1.0 ST Array 10807393 - -
          Affymetrix Rat Gene 1.0 ST Array 10807402 - -
          Affymetrix Rat Gene 1.0 ST Array 10807398 - -
          Affymetrix Rat Gene 1.0 ST Array 10807395 - -
          Affymetrix Rat Gene 1.0 ST Array 10807407 - -
          Affymetrix Rat Gene 1.0 ST Array 10807409 - -
          Affymetrix Rat Gene 1.0 ST Array 10807408 - -
          Affymetrix Rat Gene 1.0 ST Array 10807399 - -
          Affymetrix Rat Gene 1.0 ST Array 10807424 - -
          Affymetrix Rat Gene 1.0 ST Array 10807406 - -
          Affymetrix Rat Gene 1.0 ST Array 10807392 - -
          Affymetrix Rat Gene 1.0 ST Array 10807411 - -
          Affymetrix Rat Gene 1.0 ST Array 10807412 - -
          Affymetrix Rat Gene 1.1 ST Array 10807391 - -
          Rat Genome 230 2.0 Array 1388829_PM_at - -
          Rat Genome 230 2.0 Array 1388829_at - -
          Affymetrix HT Rat Focus Array 1388829_PM_at - -
          Affymetrix HT Rat Focus Array 1388829_at - -
          Affymetrix Rat Clariom S Assay PSR1900004273.rn.1 TC1900000496.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004276.rn.1 TC1900000496.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004275.rn.1 TC1900000496.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004274.rn.1 TC1900000496.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004272.rn.1 TC1900000496.rn.1 RTA TC
          Rat Expression Set 230 1388829_at - -
          Rat Expression Set 230 1388829_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807391 - -
          Rat Genome U34 Set rc_AA946348_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21413622 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357987 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline