Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01433067_m1
          See other ATG4C GE Assays ›
          Gene Symbol:
          Atg4c
          Gene Name:
          autophagy related 4C, cysteine peptidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 117862746 - 117931520 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01433067_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atg4c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107948.1 NP_001101418.1 5 - 6 1161 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001037.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          C-terminal protein lipidation
          proteolysis
          protein targeting to membrane
          autophagy
          cellular response to nitrogen starvation
          biological_process
          protein processing
          nucleophagy
          protein delipidation
          molecular_function
          cysteine-type endopeptidase activity
          peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128829 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21464 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21464 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75116 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870130 - -
          Affymetrix Rat Gene 1.0 ST Array 10870131 - -
          Affymetrix Rat Gene 1.0 ST Array 10870127 - -
          Affymetrix Rat Gene 1.0 ST Array 10870126 - -
          Affymetrix Rat Gene 1.0 ST Array 10870129 - -
          Affymetrix Rat Gene 1.0 ST Array 10870121 - -
          Affymetrix Rat Gene 1.0 ST Array 10870128 - -
          Affymetrix Rat Gene 1.0 ST Array 10870120 - -
          Affymetrix Rat Gene 1.0 ST Array 10870125 - -
          Affymetrix Rat Gene 1.0 ST Array 10870124 - -
          Affymetrix Rat Gene 1.0 ST Array 10870122 - -
          Affymetrix Rat Gene 1.0 ST Array 10870123 - -
          Affymetrix Rat Gene 1.1 ST Array 10870119 - -
          Rat Genome 230 2.0 Array 1377939_at - -
          Rat Genome 230 2.0 Array 1377939_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500006604.rn.1 TC0500001037.rn.1 RTA TC
          Rat Expression Set 230 1377939_at - -
          Rat Expression Set 230 1377939_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870119 - -
          Rat Genome U34 Set rc_AI009412_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21870827 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline