Search
Search
| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271240.1 | NP_001258169.1 | 4 - 5 | 754 | 56 | |
| XM_006233999.2 | XP_006234061.1 | 7 - 8 | 1111 | 56 | ||
| XM_006234000.2 | XP_006234062.1 | 7 - 8 | 1026 | 56 | ||
| GenBank mRNA | BC158696.1 | - | 7 - 8 | 898 | 56 | |
Set Membership: |
|