Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442148_m1
          See other FAM185A GE Assays ›
          Gene Symbol:
          Fam185a
          Gene Name:
          family with sequence similarity 185, member A
          Gene Aliases:
          RGD1562135
          Chromosome Location:
          Chr.4: 10213679 - 10269999 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442148_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam185a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128188.1 NP_001121660.1 2 - 3 769 106
          GenBank mRNA BC161871.1 - 2 - 3 769 7112030 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002230.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P760998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79780 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P760998 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P760998 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P760998 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P760998 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P563159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51484 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1396830_at - -
          Rat Genome 230 2.0 Array 1396830_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400015859.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015860.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015864.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015871.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015870.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015866.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015856.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015867.rn.1 TC0400002230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015855.rn.1 TC0400002230.rn.1 RTA TC
          Rat Expression Set 230 1396830_PM_at - -
          Rat Expression Set 230 1396830_at - -
          Rat Genome U34 Set rc_AI231011_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358641 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline