Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442238_m1
          See other DNAJC2 GE Assays ›
          Gene Symbol:
          Dnajc2
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 2
          Gene Aliases:
          MIDA1, Zrf2
          Chromosome Location:
          Chr.4: 9881513 - 9908693 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442238_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053776.2 NP_446228.2 7 - 8 862 172
          XM_008762628.1 XP_008760850.1 8 - 9 1171 172
          XM_008762629.1 XP_008760851.1 5 - 6 424 172
          GenBank mRNA AF118853.1 - 7 - 8 835 172
          AY322161.1 - 7 - 8 835 172
          AY322162.1 - 7 - 8 835 172
          AY322163.1 - 7 - 8 746 172
          BC088838.1 - 7 - 8 862 7132881 172

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000155.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajc2

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          transcription, DNA-templated
          chromatin modification
          negative regulation of cell growth
          positive regulation of transcription, DNA-templated
          negative regulation of DNA biosynthetic process
          DNA binding
          chromatin binding
          Hsp70 protein binding
          histone binding
          ubiquitin binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33362 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023599 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023599 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P367499 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701255720 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69066 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99677 - -
          Agilent Rat cDNA Microarray (G4105A) 701255720 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853111 - -
          Affymetrix Rat Gene 1.0 ST Array 10853119 - -
          Affymetrix Rat Gene 1.0 ST Array 10853114 - -
          Affymetrix Rat Gene 1.0 ST Array 10853128 - -
          Affymetrix Rat Gene 1.0 ST Array 10853127 - -
          Affymetrix Rat Gene 1.0 ST Array 10853122 - -
          Affymetrix Rat Gene 1.0 ST Array 10853121 - -
          Affymetrix Rat Gene 1.0 ST Array 10853112 - -
          Affymetrix Rat Gene 1.0 ST Array 10853113 - -
          Affymetrix Rat Gene 1.0 ST Array 10853115 - -
          Affymetrix Rat Gene 1.0 ST Array 10853129 - -
          Affymetrix Rat Gene 1.0 ST Array 10853117 - -
          Affymetrix Rat Gene 1.0 ST Array 10853120 - -
          Affymetrix Rat Gene 1.0 ST Array 10853118 - -
          Affymetrix Rat Gene 1.0 ST Array 10853116 - -
          Affymetrix Rat Gene 1.0 ST Array 10853124 - -
          Affymetrix Rat Gene 1.0 ST Array 10853125 - -
          Affymetrix Rat Gene 1.0 ST Array 10853123 - -
          Affymetrix Rat Gene 1.0 ST Array 10853126 - -
          Affymetrix Rat Gene 1.1 ST Array 10853110 - -
          Rat Genome 230 2.0 Array 1391487_at - -
          Rat Genome 230 2.0 Array 1373004_PM_at - -
          Rat Genome 230 2.0 Array 1373004_at - -
          Rat Genome 230 2.0 Array 1391487_PM_at - -
          Affymetrix HT Rat Focus Array 1391487_PM_at - -
          Affymetrix HT Rat Focus Array 1373004_at - -
          Affymetrix HT Rat Focus Array 1373004_PM_at - -
          Affymetrix HT Rat Focus Array 1391487_at - -
          Affymetrix Rat Clariom S Assay PSR0400001195.rn.1 TC0400000155.rn.1 RTA TC
          Rat Expression Set 230 1373004_at - -
          Rat Expression Set 230 1373004_PM_at - -
          Rat Expression Set 230 1391487_PM_at - -
          Rat Expression Set 230 1391487_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853110 - -
          Rat Genome U34 Set rc_AI044807_at - -
          Rat Genome U34 Set rc_AA964764_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22404459 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369836 - -
          RatRef-12 Expression BeadChip ILMN_55430 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline