Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442797_m1
          See other GDF6 GE Assays ›
          Gene Symbol:
          Gdf6
          Gene Name:
          growth differentiation factor 6
          Gene Aliases:
          BMP-13, gdf16
          Chromosome Location:
          Chr.5: 23056345 - 23072666 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442797_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gdf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013038.1 NP_001013056.1 1 - 2 523 109
          GenBank mRNA AJ537426.2 - 1 - 2 523 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000184.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gdf6

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          positive regulation of pathway-restricted SMAD protein phosphorylation
          BMP signaling pathway
          activin receptor signaling pathway
          growth
          regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          cell development
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          retinal cell apoptotic process
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334900 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334900 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P334900 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334900 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867594 - -
          Affymetrix Rat Gene 1.0 ST Array 10867595 - -
          Affymetrix Rat Gene 1.1 ST Array 10867593 - -
          Rat Genome 230 2.0 Array 1391038_at - -
          Rat Genome 230 2.0 Array 1391038_PM_at - -
          Affymetrix HT Rat Focus Array 1391038_at - -
          Affymetrix HT Rat Focus Array 1391038_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500001193.rn.1 TC0500000184.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500001192.rn.1 TC0500000184.rn.1 RTA TC
          Rat Expression Set 230 1391038_at - -
          Rat Expression Set 230 1391038_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867593 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21706401 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350352 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline