Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443731_g1
          See other PES1 GE Assays ›
          Gene Symbol:
          Pes1
          Gene Name:
          pescadillo ribosomal biogenesis factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 84211600 - 84227980 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443731_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pes1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044228.1 NP_001037693.1 13 - 14 1690 55
          GenBank mRNA BC105910.1 - 13 - 14 1690 7388101 55

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          maturation of LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          maturation of 5.8S rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          rRNA processing
          nucleolus organization
          cell proliferation
          protein localization to organelle
          ribosome biogenesis
          regulation of cell cycle
          RNA binding
          ribonucleoprotein complex binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35672 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005313 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005313 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P478074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73471 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9366 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773770 - -
          Affymetrix Rat Gene 1.0 ST Array 10773763 - -
          Affymetrix Rat Gene 1.0 ST Array 10773772 - -
          Affymetrix Rat Gene 1.0 ST Array 10773769 - -
          Affymetrix Rat Gene 1.0 ST Array 10773766 - -
          Affymetrix Rat Gene 1.0 ST Array 10773768 - -
          Affymetrix Rat Gene 1.0 ST Array 10773767 - -
          Affymetrix Rat Gene 1.0 ST Array 10773762 - -
          Affymetrix Rat Gene 1.0 ST Array 10773771 - -
          Affymetrix Rat Gene 1.0 ST Array 10773764 - -
          Affymetrix Rat Gene 1.0 ST Array 10773765 - -
          Affymetrix Rat Gene 1.0 ST Array 10773761 - -
          Affymetrix Rat Gene 1.1 ST Array 10773760 - -
          Rat Genome 230 2.0 Array 1389238_PM_at - -
          Rat Genome 230 2.0 Array 1389238_at - -
          Affymetrix HT Rat Focus Array 1389238_at - -
          Affymetrix HT Rat Focus Array 1389238_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400006358.rn.1 TC1400000889.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400006359.rn.1 TC1400000889.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400006357.rn.1 TC1400000889.rn.1 RTA TC
          Rat Expression Set 230 1389238_PM_at - -
          Rat Expression Set 230 1389238_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773760 - -
          Rat Genome U34 Set rc_AA955980_g_at - -
          Rat Genome U34 Set rc_AA955980_at - -
          Rat Genome U34 Set rc_AI235689_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21668493 - -
          ABI Rat Genome Survey Microarray 21758106 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364114 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline