Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447193_m1
          See other HMGN3 GE Assays ›
          Gene Symbol:
          Hmgn3
          Gene Name:
          high mobility group nucleosomal binding domain 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 90628203 - 90665059 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447193_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmgn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007020.2 NP_001007021.1 4 - 5 280 74
          NM_001201353.1 NP_001188282.1 4 - 5 280 74
          NM_001201354.1 NP_001188283.1 4 - 5 280 74
          NM_001201355.1 NP_001188284.1 4 - 5 280 74
          XM_006243445.2 XP_006243507.1 4 - 5 785 74
          XM_006243446.2 XP_006243508.1 4 - 5 785 74
          XM_008766200.1 XP_008764422.1 4 - 5 785 74
          GenBank mRNA BC082036.1 - 4 - 5 235 7130894 74
          CB751211.1 - 4 - 5 163 74
          CB774488.1 - 4 - 5 248 74
          FM058030.1 - 4 - 5 280 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003031.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmgn3

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          chromatin modification
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of insulin secretion involved in cellular response to glucose stimulus
          chromatin binding
          nucleosomal DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P156682 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P518462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155241 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P518462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P518462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156682 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P156682 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P518462 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P518462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55991 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103207 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919013 - -
          Affymetrix Rat Gene 1.0 ST Array 10919011 - -
          Affymetrix Rat Gene 1.0 ST Array 10919015 - -
          Affymetrix Rat Gene 1.0 ST Array 10919012 - -
          Affymetrix Rat Gene 1.0 ST Array 10919014 - -
          Affymetrix Rat Gene 1.0 ST Array 10919016 - -
          Affymetrix Rat Gene 1.1 ST Array 10919010 - -
          Rat Genome 230 2.0 Array 1371989_PM_at - -
          Rat Genome 230 2.0 Array 1371989_at - -
          Affymetrix HT Rat Focus Array 1371989_PM_at - -
          Affymetrix HT Rat Focus Array 1371989_at - -
          Affymetrix Rat Clariom S Assay PSR0800024256.rn.1 TC0800003031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800024255.rn.1 TC0800003031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800024259.rn.1 TC0800003031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800024253.rn.1 TC0800003031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800024257.rn.1 TC0800003031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800024254.rn.1 TC0800003031.rn.1 RTA TC
          Rat Expression Set 230 1371989_PM_at - -
          Rat Expression Set 230 1371989_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919010 - -
          Rat Genome U34 Set rc_AI178675_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20864587 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362726 - -
          RatRef-12 Expression BeadChip ILMN_68964 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline