Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449463_m1
          See other UPP2 GE Assays ›
          Gene Symbol:
          Upp2
          Gene Name:
          uridine phosphorylase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 44806106 - 44846464 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449463_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Upp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106481.1 NP_001099951.1 6 - 7 1224 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000638.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide catabolic process
          UMP salvage
          uridine phosphorylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P192224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192224 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192224 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P192224 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192224 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29068 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836318 - -
          Affymetrix Rat Gene 1.0 ST Array 10836313 - -
          Affymetrix Rat Gene 1.0 ST Array 10836315 - -
          Affymetrix Rat Gene 1.0 ST Array 10836317 - -
          Affymetrix Rat Gene 1.0 ST Array 10836316 - -
          Affymetrix Rat Gene 1.0 ST Array 10836319 - -
          Affymetrix Rat Gene 1.0 ST Array 10836320 - -
          Affymetrix Rat Gene 1.1 ST Array 10836312 - -
          Affymetrix Rat Clariom S Assay PSR0300004856.rn.1 TC0300000638.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10836312 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20743320 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline