Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449467_m1
          See other RND3 GE Assays ›
          Gene Symbol:
          Rnd3
          Gene Name:
          Rho family GTPase 3
          Gene Aliases:
          Arhe, RHOE
          Chromosome Location:
          Chr.3: 36643374 - 36660727 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449467_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007641.1 NP_001007642.1 3 - 4 536 100
          GenBank mRNA AY461427.1 - 3 - 4 536 100
          BC168142.1 - 564 9100645 100
          FQ210593.1 - 575 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003100.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnd3

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1050 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P414460 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P414460 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P414460 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P414460 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P429083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845126 - -
          Affymetrix Rat Gene 1.0 ST Array 10845125 - -
          Affymetrix Rat Gene 1.0 ST Array 10845128 - -
          Affymetrix Rat Gene 1.0 ST Array 10845127 - -
          Affymetrix Rat Gene 1.0 ST Array 10845129 - -
          Affymetrix Rat Gene 1.1 ST Array 10845124 - -
          Rat Genome 230 2.0 Array 1377663_PM_at - -
          Rat Genome 230 2.0 Array 1386062_at - -
          Rat Genome 230 2.0 Array 1376066_at - -
          Rat Genome 230 2.0 Array 1394077_at - -
          Rat Genome 230 2.0 Array 1376066_PM_at - -
          Rat Genome 230 2.0 Array 1386062_PM_at - -
          Rat Genome 230 2.0 Array 1377663_at - -
          Rat Genome 230 2.0 Array 1394077_PM_at - -
          Affymetrix HT Rat Focus Array 1377663_at - -
          Affymetrix HT Rat Focus Array 1394077_PM_at - -
          Affymetrix HT Rat Focus Array 1394077_at - -
          Affymetrix HT Rat Focus Array 1386062_at - -
          Affymetrix HT Rat Focus Array 1386062_PM_at - -
          Affymetrix HT Rat Focus Array 1376066_PM_at - -
          Affymetrix HT Rat Focus Array 1376066_at - -
          Affymetrix HT Rat Focus Array 1377663_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300023864.rn.1 TC0300003100.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300023866.rn.1 TC0300003100.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300023863.rn.1 TC0300003100.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300023865.rn.1 TC0300003100.rn.1 RTA TC
          Rat Expression Set 230 1376066_PM_at - -
          Rat Expression Set 230 1376066_at - -
          Rat Expression Set 230 1394077_at - -
          Rat Expression Set 230 1386062_PM_at - -
          Rat Expression Set 230 1377663_PM_at - -
          Rat Expression Set 230 1394077_PM_at - -
          Rat Expression Set 230 1386062_at - -
          Rat Expression Set 230 1377663_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845124 - -
          Rat Genome U34 Set rc_AA859776_at - -
          Rat Genome U34 Set rc_AI103572_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21561199 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376851 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline