Search
Search
| Gene Symbol: Asgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012503.2 | NP_036635.1 | 3 - 4 | 248 | 56 | |
| XM_006246579.1 | XP_006246641.1 | 3 - 4 | 360 | 56 | ||
| XM_008767768.1 | XP_008765990.1 | 3 - 4 | 380 | 56 | ||
| XM_008767769.1 | XP_008765991.1 | 3 - 4 | 324 | 56 | ||
| GenBank mRNA | BC088154.1 | - | 3 - 4 | 248 | 7372227 | 56 |
| FQ209763.1 | - | 3 - 4 | 284 | 56 | ||
| FQ218644.1 | - | 3 - 4 | 284 | 56 | ||
| FQ218824.1 | - | 3 - 4 | 284 | 56 | ||
| FQ219107.1 | - | 3 - 4 | 321 | 56 | ||
| FQ219710.1 | - | 3 - 4 | 365 | 56 | ||
| M21770.1 | - | 2 - 3 | 150 | 56 | ||
Set Membership: |
|