Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455353_m1
          See other IQGAP3 GE Assays ›
          Gene Symbol:
          Iqgap3
          Gene Name:
          IQ motif containing GTPase activating protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 187447501 - 187489630 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455353_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Iqgap3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191709.1 NP_001178638.1 5 - 6 433 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001527.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          activation of MAPK activity
          positive regulation of protein phosphorylation
          Ras protein signal transduction
          regulation of cell size
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of mammary gland epithelial cell proliferation
          ERK1 and ERK2 cascade
          cellular response to organic substance
          calmodulin binding
          Ras GTPase binding
          Rho GTPase binding
          myosin VI light chain binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P182546 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P182546 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P182546 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154297 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P182546 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P154297 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482880 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1374902_at - -
          Rat Genome 230 2.0 Array 1374902_PM_at - -
          Affymetrix HT Rat Focus Array 1374902_at - -
          Affymetrix HT Rat Focus Array 1374902_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200009550.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009588.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009592.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009593.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009587.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009558.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009574.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009578.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009549.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009548.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009561.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009563.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009595.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009559.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009586.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009571.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009590.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009556.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009585.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009557.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009577.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009582.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009594.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009547.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009567.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009568.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009551.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009566.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009580.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009565.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009564.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009583.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009573.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009581.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009560.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009553.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009572.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009576.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009555.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009569.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009570.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009554.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009552.rn.1 TC0200001527.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009589.rn.1 TC0200001527.rn.1 RTA TC
          Rat Expression Set 230 1374902_PM_at - -
          Rat Expression Set 230 1374902_at - -
          Rat Genome U34 Set rc_AI010211_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361765 - -
          RatRef-12 Expression BeadChip ILMN_62394 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline