Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455365_g1
          See other GPATCH4 GE Assays ›
          Gene Symbol:
          Gpatch4
          Gene Name:
          G patch domain containing 4
          Gene Aliases:
          RGD1311086
          Chromosome Location:
          Chr.2: 187404897 - 187424047 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455365_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpatch4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024979.1 NP_001020150.1 7 - 8 596 102
          XM_008761129.1 XP_008759351.1 8 - 9 1127 102
          XM_008761130.1 XP_008759352.1 8 - 9 1177 102
          XM_008761131.1 XP_008759353.1 7 - 8 1113 102
          XM_008761132.1 XP_008759354.1 8 - 9 1291 102
          GenBank mRNA BC093381.1 - 7 - 8 596 7388663 102
          FQ233538.1 - 4 - 5 318 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001526.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpatch4

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          nucleic acid binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P149662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137436 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P149662 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P149662 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P149662 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P149662 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10884468 - -
          Affymetrix Rat Gene 1.0 ST Array 10816427 - -
          Affymetrix Rat Gene 1.0 ST Array 10816430 - -
          Affymetrix Rat Gene 1.0 ST Array 10816424 - -
          Affymetrix Rat Gene 1.0 ST Array 10816419 - -
          Affymetrix Rat Gene 1.0 ST Array 10816431 - -
          Affymetrix Rat Gene 1.0 ST Array 10816428 - -
          Affymetrix Rat Gene 1.0 ST Array 10816422 - -
          Affymetrix Rat Gene 1.0 ST Array 10816432 - -
          Affymetrix Rat Gene 1.0 ST Array 10816426 - -
          Affymetrix Rat Gene 1.0 ST Array 10816423 - -
          Affymetrix Rat Gene 1.0 ST Array 10816421 - -
          Affymetrix Rat Gene 1.0 ST Array 10816429 - -
          Affymetrix Rat Gene 1.0 ST Array 10816425 - -
          Affymetrix Rat Gene 1.0 ST Array 10816420 - -
          Affymetrix Rat Gene 1.1 ST Array 10884466 - -
          Affymetrix Rat Gene 1.1 ST Array 10816418 - -
          Rat Genome 230 2.0 Array 1393128_PM_at - -
          Rat Genome 230 2.0 Array 1382187_PM_at - -
          Rat Genome 230 2.0 Array 1382186_PM_a_at - -
          Rat Genome 230 2.0 Array 1382188_PM_at - -
          Rat Genome 230 2.0 Array 1393128_at - -
          Rat Genome 230 2.0 Array 1382187_at - -
          Rat Genome 230 2.0 Array 1382186_a_at - -
          Rat Genome 230 2.0 Array 1382188_at - -
          Affymetrix HT Rat Focus Array 1393128_PM_at - -
          Affymetrix HT Rat Focus Array 1382186_PM_a_at - -
          Affymetrix HT Rat Focus Array 1382186_a_at - -
          Affymetrix HT Rat Focus Array 1382187_PM_at - -
          Affymetrix HT Rat Focus Array 1382188_PM_at - -
          Affymetrix HT Rat Focus Array 1382188_at - -
          Affymetrix HT Rat Focus Array 1393128_at - -
          Affymetrix HT Rat Focus Array 1382187_at - -
          Affymetrix Rat Clariom S Assay PSR0200009544.rn.1 TC0200001526.rn.1 RTA TC
          Rat Expression Set 230 1382187_PM_at - -
          Rat Expression Set 230 1382187_at - -
          Rat Expression Set 230 1382188_at - -
          Rat Expression Set 230 1382188_PM_at - -
          Rat Expression Set 230 1382186_a_at - -
          Rat Expression Set 230 1393128_PM_at - -
          Rat Expression Set 230 1382186_PM_a_at - -
          Rat Expression Set 230 1393128_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816418 - -
          Rat Genome U34 Set rc_AI029955_at - -
          Rat Genome U34 Set rc_AI136314_at - -
          Rat Genome U34 Set rc_AI072485_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21085652 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650908 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline