Search
Search
| Gene Symbol: Casc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106627.3 | NP_001100097.2 | 7 - 8 | 631 | 61 | |
| XM_008763378.1 | XP_008761600.1 | 3 - 4 | 1068 | 61 | ||
| GenBank mRNA | BC160907.1 | - | 7 - 8 | 631 | 7455489 | 61 |
| CV108923.1 | - | 7 - 8 | 630 | 61 | ||
Set Membership: |
|